物質
copper
包装
pack of 1 unit
メーカー/製品名
USB-C Cable Sirius
テクニック
: suitable using airmonitoring
適合性
MAS-100® Sirius®
関連するカテゴリー
詳細
The USB-C Cable is included with the MAS-100 Sirius but can also be ordered separately.
This cable is identified as USB Type C 2.0 PD fast charging, supporting up to 60W (20V, 3A) and has a length of 1.5 meters.
This cable is identified as USB Type C 2.0 PD fast charging, supporting up to 60W (20V, 3A) and has a length of 1.5 meters.
アプリケーション
The USB-C cable is used for charging as well as for connecting to a PC to access the instrument-integrated software via a browser.
法的情報
MAS-100 is a registered trademark of MBV AG
MAS-100 Sirius is a registered trademark of MBV AG