コンテンツへスキップ
Merck
  • Matrix Metalloproteinase-9 (279R/Q) Polymorphism is Associated with Clinical Severity and Airflow Limitation in Tunisian Patients with Chronic Obstructive Pulmonary Disease.
Matrix Metalloproteinase-9 (279R/Q) Polymorphism is Associated with Clinical Severity and Airflow Limitation in Tunisian Patients with Chronic Obstructive Pulmonary Disease.
Molecular diagnosis & therapy (2015-08-26)
Sarra Bchir, Hela Ben Nasr, Imen Rekik Hakim, Amel Ben Anes, Saloua Yacoub, Abdelhamid Garrouch, Mohamed Benzarti, Brigitte Bauvois, Zouhair Tabka, Karim Chahed
要旨
The aim of this study was to investigate the role of matrix metalloproteinase-9 (MMP-9) C-1562T and 279R/Q (836G>A) polymorphisms in the development of chronic obstructive pulmonary disease (COPD) in Tunisians and to determine their impact on disease progression and airflow obstruction. Pulmonary functional tests were evaluated by body plethysmography. MMP-9 genotypes were determined in patients with COPD (n = 138) and healthy controls (n = 216) by polymerase chain reaction-restriction fragment length polymorphism (PCR-RFLP). Serum MMP-9 and interleukin-6 (IL-6) levels were determined by enzyme-linked immunosorbent assays (ELISA) and activity of MMP-9 was evaluated by gelatin zymography. No significant association was found between genetic variations in MMP-9 C-1562T and 279R/Q polymorphisms and the risk of development of COPD. However, a significant correlation was retrieved between the 279 R/Q polymorphism and disease severity (P = 0.02). In addition, homozygous Q (A) genotype was associated with a poorer lung function with a fall in forced expiratory volume in 1 s (FEV1) (%) and forced vital capacity (FVC%) among COPD patients compared with both AG and GG individuals (52.06 ± 19.6 vs. 59.08 ± 17.19, P = 0.03 and 72.41 ± 21.42 vs. 82.98 ± 16.48, P = 0.002, respectively). Using ELISA, a higher level of MMP-9 was found in patients with the CT genotype (P = 0.03), while no significant impact of the 279R/Q polymorphism was observed (P = 0.48). In contrast, by using zymography gel analysis, MMP-9 activity was enhanced in individuals carrying the R(G) allele in comparison with those homozygous for the Q(A) variant (P = 0.02). Our results support a role for the 279R/Q polymorphism in physiological alterations that may affect progression and severity of COPD. These findings could be related to the decreased activity of MMP-9 among COPD patients carrying the 279Q variant.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
塩化マグネシウム 溶液, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
アジ化ナトリウム, ReagentPlus®, ≥99.5%
Sigma-Aldrich
塩化ナトリウム, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
塩化マグネシウム, ≥98%
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
塩化ナトリウム, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
塩化カルシウム 溶液, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
塩化ナトリウム, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
塩化ナトリウム 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
塩化カルシウム, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
アジ化ナトリウム, BioUltra, ≥99.5% (T)
Sigma-Aldrich
塩化ナトリウム 溶液, 5 M
Sigma-Aldrich
塩化マグネシウム, powder, <200 μm
Sigma-Aldrich
塩化ナトリウム, JIS special grade, ≥99.5%
Sigma-Aldrich
アジ化ナトリウム, purum p.a., ≥99.0% (T)
Sigma-Aldrich
塩化ナトリウム, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
塩化マグネシウム 溶液, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
アジ化ナトリウム, BioXtra
Sigma-Aldrich
塩化ナトリウム, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
塩化ナトリウム 溶液, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
塩化ナトリウム, 99.999% trace metals basis
Sigma-Aldrich
塩化カルシウム, powder, 99.99% trace metals basis
Sigma-Aldrich
塩化マグネシウム 溶液, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
塩化マグネシウム, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
塩化マグネシウム 溶液, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
塩化カルシウム, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
塩化ナトリウム, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
塩化ナトリウム 溶液, 0.85%