Thermal Transitions of Homopolymers: Glass Transition & Melting Point
Literature values for the glass transition temperature, (Tg), and melting temperature, (Tm) for the more common homopolymers are listed in the table below. Polymers are listed by the repeating unit in the polymer chain. The polymers and corresponding monomers listed are available from us. Literature values for a given material can vary widely. The values reported have been obtained from various sources and represent the most commonly reported numbers.1 Several factors can influence the reported values, including molecular weight, molecular weight distribution, tacticity, thermal history, purity, and method of measurement.
Thermal Transitions of Homopolymers: Glass Transition (Tg) & Melting Point (Tm) Temperatures
| Repeating Unit | Tg (°C) | Tm (°C) | Repeating Unit | Tg (°C) | Tm (°C) |
|---|---|---|---|---|---|
| Acenaphthylene | 214 | N,N-Dimethylacrylamide | 89 | ||
| Acetaldehyde | -32 | 165 | Dimethylaminoethyl methacrylate | 19 | |
| 4-Acetoxystyrene | 116 | 2,6-Dimethyl-1,4-phenylene oxide | 167 | ||
| Acrylamide | 165 | Dimethylsiloxane | -127 | -40 | |
| Acrylic acid | 105 | 2,4-Dimethylstyrene | 112 | ||
| Acrylonitrile, syndiotactic | 125 | 319 | 2,5-Dimethylstyrene | 143 | |
| Allyl glycidyl ether | -78 | 3,5-Dimethylstyrene | 104 | ||
| Benzyl acrylate | 6 | Dodecyl acrylate | -3 | ||
| Benzyl methacrylate | 54 | Dodecyl methacrylate | -65 | ||
| Bisphenol A-alt-epichlorohydrin | 100 | Dodecyl vinyl ether | -62 | ||
| Bisphenol A terephthalate | 205 | Epibromohydrin | -14 | ||
| Bisphenol carbonate | 174 | Epichlorohydrin | -22 | ||
| Bisphenol F carbonate | 147 | 1,2-Epoxybutane | -70 | ||
| Bisphenol Z carbonate | 175 | 1,2-Epoxydecane | -70 | ||
| 4-Bromostyrene | 118 | 1,2-Epoxyoctane | -67 | ||
| cis-Butadiene | 102 | 1 | 2-Ethoxyethyl acrylate | -50 | |
| trans-Butadiene | -58 | 148 | 4-Ethoxystyrene | 86 | |
| 1-Butene | -24 | 171 | Ethyl acrylate | -24 | |
| N-tert-Butylacrylamide | 128 | Ethyl cellulose | 43 | ||
| Butyl acrylate | -54 | Ethylene, HDPE | -125 | 130 | |
| sec-Butyl acrylate | -26 | Ethylene adipate | -45 | 54 | |
| tert-Butyl acrylate | 43-107 | 193 | Ethylene-trans-1,4-cyclohexyldicarboxylate | 18 | – |
| 2-tert-Butylaminoethyl methacrylate | 33 | Ethylene isophthatate | 51 | ||
| Butyl glydicyl ether | -79 | Ethylene malonate | -29 | ||
| Butyl methacrylate | 20 | Ethylene 2,6-napthalenedicarboxylate | 113 | ||
| tert-Butyl methacrylate | 118 | Ethylene oxide | -66 | 66 | |
| 4-tert-Butylstyrene | 127 | Ethylene terephthalate | 72 | 265 | |
| tert-Butyl vinyl ether | 88 | 250 | 2-Ethylhexyl acrylate | -50 | |
| Butyl vinyl ether | -55 | 64 | 2-Ethylhexyl methacrylate | -10 | |
| ε-Caprolactone | -60 | 2-Ethylhexyl vinyl ether | -66 | ||
| Cellulose nitrate | 53 | Ethyl methacrylate | 65 | ||
| Cellulose tripropionate | Ethyl vinyl ether | -43 | 86 | ||
| cis-Chlorobutadiene | -20 | 80 | 4-Fluorostyrene | 95 | |
| trans-Chlorobutadiene | -40 | 101 | Formaldehyde | -82 | 181 |
| 2-Chlorostyrene | 119 | Hexadecyl acrylate | 35 | ||
| 3-Chlorostyrene | 90 | Hexadecyl methacrylate | 15 | ||
| 4-Chlorostyrene | 110 | Hexyl acrylate | -57 | ||
| Chlorotrifluoroethylene | 52 | 214 | Hexyl methacrylate | -5 | |
| 2-Cyanoethyl acrylate | 4 | 2-Hydropropyl methacrylate | 76 | ||
| Cyclohexyl acrylate | 19 | Hydroquinone-alt-epichlorohydrin | 60 | ||
| Cyclohexyl methacrylate | 92 | 2-Hydroxyethyl methacrylate | 57 | ||
| Cyclohexyl vinyl ether | 81 | Indene | 85 | ||
| 2,6-Dichlorostyrene | 167 | Isobornyl acrylate | 94 | ||
| Diethylaminoethyl methacrylate | 20 | Isobornyl methacrylate | 110 | ||
| Isobutyl acrylate | -24 | p-Phenylene terephthalamide | 345 | ||
| Isobutylene | -73 | Phenylene vinylene | 80 | 380 | |
| Isobutyl methacrylate | 53 | Phenyl methacrylate | 110 | ||
| Isobutyl vinyl ether | -19 | 165 | Phenyl vinyl ketone | 74 | |
| cis-Isoprene | -63 | 28 | Potassium acrylate | 194 | |
| trans-Isoprene | -66 | 65 | Propylene, atactic | -13 | |
| N-Isopropylacrylamide | 85-130 | Propylene, isotactic | -8 | 176 | |
| Isopropyl acrylate, isotactic | -11 | 162 | Propylene, syndiotactic | -8 | |
| Isopropyl methacrylate | 81 | Propylene oxide | -75 | 66 | |
| Methacrylic acid | 228 | Propyl vinyl ether | -49 | 76 | |
| Methacrylic anhydride | 159 | Sodium acrylate | 230 | ||
| Methacrylonitrile | 120 | Sodium methacrylate | 310 | ||
| 2-Methoxyethyl acrylate | -50 | Styrene, atactic | 100 | ||
| 4-Methoxystyrene | 113 | Styrene, isotactic | 100 | 240 | |
| Methyl acrylate | 10 | Tetrabromobisphenol A carbonate | 157 | ||
| Methyl cellulose | Tetrafluoroethylene | 117 | 327 | ||
| Methyl glycidyl ether | -62 | Tetrahydrofuran | -84 | ||
| Methyl methacrylate, atactic | 105,120 | Tetramethylene adipate | -118 | ||
| Methyl methacrylate, syndiotactic | 115 | 200 | Tetramethylene terephthalate | 17 | 232 |
| 4-Methylpentene | 29 | 250 | Thio-1,4-phenylene | 97 | 285 |
| Methylphenylsiloxane | -86 | 2,2,2-Trifluoroethyl acrylate | -10 | ||
| Methylstyrene | 20 | Trimethylene oxide | -78 | ||
| 3-Methylstyrene | 97 | Trimethylsilyl methacrylate | 68 | ||
| 4-Methylstyrene | 97 | 2,4,6-Trimethylstyrene | 162 | ||
| Methyl vinyl ether | -31 | 144 | Vinyl acetal | 355 | 82 |
| Nylon 4,6 (tetramethylene adipamide) | 43 | Vinyl acetate | 30 | ||
| Nylon 6 (-caprolactam) | 52 | 225 | Vinyl alcohol | 85 | 220 |
| Nylon 6,6 (hexamethylene adipamide) | 50 | 265 | Vinyl benzoate | 71 | |
| Nylon 6,9 (hexamethylene azelamide) | 58 | Vinyl 4-tert-butylbenzoate | 101 | – | |
| Nylon 6,10 (hexamethylene sebacamide) | 50 | 227 | Vinyl butyral | 322 | 49 |
| Nylon 6,12 (hexamethylene dodecane-diamide) | 46 | Vinyl carbazole | 227 | 320 | |
| Nylon 11 (ω−undecanamide) | 42 | 189 | Vinyl chloride | 81 | 227 |
| Nylon 12 (ω-dodecanamide) | 41 | 179 | Vinyl cyclohexanoate | 76 | |
| 1-Octadecene | 55 | Vinylferrocene | 189 | ||
| Octadecyl methacrylate | -100 | Vinyl fluoride | 41 | 200 | |
| 1-Octene | -63 | Vinyl formal | 105 | ||
| Octyl methacrylate | -20 | Vinylidene chloride | -18 | 200 | |
| Oxy-4,4'-biphenyleneoxy-1,4-phenylenesulfonyl-1,4-phenylene | 230 | 290 | Vinylidene fluoride | -40 | 171 |
| Oxy-1,4-phenylenesulfonyl-1,4-phenyleneoxy-1,4-phenyleneisopropylidene-1,4-phenylene | 165 | 190 | 2-Vinyl naphthalene | 151 | |
| Oxy-1,4-phenylenesulfonyl-1,4-phenylene ether | 214 | 230 | Vinyl pivalate | 86 | |
| p-Phenylene isophthalamide | 225 | 380 | Vinyl propionate | 10 | |
| 2-Vinylpyridine | 104 | ||||
| 4-Vinylpyridine | 142 | ||||
| 1-Vinyl-2-pyrrolidone | 54 | ||||
| Vinyl trifluoroacetate | 46 |