Merck
Physicochemical determinants of human renal clearance.
Journal of medicinal chemistry (2009-05-19)
Manthena V S Varma, Bo Feng, R Scott Obach, Matthew D Troutman, Jonathan Chupka, Howard R Miller, Ayman El-Kattan
ABSTRAKT
Kidney plays an important role in the elimination of drugs, especially with low or negligible hepatic clearance. An analysis of the interrelation of physicochemical properties and the human renal clearance for a data set of 391 drugs or compounds tested in humans is presented. The data set indicated that lipophilicity shows a negative relationship while polar descriptors show a positive relationship with renal clearance. Analysis of net secreted and net reabsorbed subsets revealed that hydrophilic ionized compounds are probable compounds to show net secretion and a possible drug-drug interaction due to their likely interaction with uptake transporters and inherent low passive reabsorption. The physicochemical space and renal clearance were also statistically analyzed by therapeutic area. In conclusion, ionization state, lipophilicity, and polar descriptors are found to be the physicochemical determinants of renal clearance. These fundamental properties can be valuable in early prediction of human renal clearance and can aid the chemist in structural modifications to optimize drug disposition.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Ibuprofen
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Cyclosporin A, VETRANAL®, analytical standard
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Nitrofurantoin, 98.0-102.0% (EP, UV), meets EP testing specifications
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Sigma-Aldrich
Mevinolin from Aspergillus sp., ≥98% (HPLC)
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Sigma-Aldrich
Prednisone, ≥98%
Sigma-Aldrich
Chlorambucil
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, Molecular Biology, ≥95%
Sigma-Aldrich
Glipizide, solid
Sigma-Aldrich
Oxazepam
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Pindolol, ≥98% (TLC), powder
Sigma-Aldrich
Prednisolone, ≥99%
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sigma-Aldrich
(±)-Sulpiride