Merck
[Genetic transformation of Pinellia ternata with Agrobacterium tumefaciens-mediated sHSP genes].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-05-01)
Zhao-Yang Guo, Ting-Ting Cui, Jian-Ping Xue, Yan-Fang Zhu, Ai-Min Zhang, Wei Sheng, Jing-Tong Teng
ABSTRAKT
To establish an efficient genetic transformation system of Pinellia ternata. With petioles from test-tube seedlings of P. ternata as explants, Agrobacterium tumefaciens mediation method was adopted to explore the effect of phenolic substances, A. tumefaciens's concentration, infection time, pre-incubation time and co-cultivation time on genetic transformation efficiency of P. ternata. The genetic transformation efficiency could be effectively enhanced by infecting in A. tumefaciens culture containing AS 40 mg x L(-1) for 15 min for three days. The petioles were put into the differentiation medium containing 150 mg x L(-1) Kan and 350 mg x L(-1) Carb to screening and cultivation. After around 30 days, microtubers could be observed at both sides of the petioles. Gus staining and PCR verification on the regenerated plants showed that the exogenous gene sHSP had been integrated into genome of P. ternata.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
β-Glucuronidase from limpets (Patella vulgata), Type L-II, lyophilized powder, 1,000,000-3,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-5, lyophilized powder, ≥400,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-1, partially purified powder, ≥300,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type HP-2S, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-3AF, aqueous solution, ≥60,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-3, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-2, aqueous solution, ≥85,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type HP-2, aqueous solution, ≥100,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, ≥10,000,000 units/g protein (30 min assay), recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type IX-A, lyophilized powder, 1,000,000-5,000,000 units/g protein (30 min assay)
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, aqueous glycerol solution, ≥5,000,000 units/g protein, pH 6.8 (biuret)
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type VII-A, lyophilized powder, 5,000,000-20,000,000 units/g protein, pH 6.8 (30 min assay)
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-3, ≥2,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-1, ≥1,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix aspersa (garden snail), Type HA-4
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, ≥20,000 units/mg protein, recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, >20,000,000 units/g protein, recombinant, expressed in E. coli, aqueous glycerol solution