Merck
[Effect of cordyceps polysaccharide on lipid peroxidation of rats with dimethylnitrosamine-induced liver fibrosis].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-05-15)
Jing-Hua Peng, Xue-Mei Li, Yi-Yang Hu, Qin Feng
ABSTRAKT
To observe the pharmacological effect of Cordyceps polysaccharide on dimethylnitrosamine (DMN)-induced liver fibrosis in rats. DMN rat liver fibrosis model was established and divided into the normal group (N, n = 6), the model group (M, n = 11), the Cordyceps polysaccharide group (C, n = 8) and the colchicine group (Q, n = 9). During the modeling for four weeks, Cordyceps polysaccharide (60 mg x kg(-1)) and colchicine (0.1 mg x kg(-1)) were orally administered for three weeks, while the model and normal groups were given disinfected water of the same amount. serum ALT, AST, GGT and Alb, TBil content; content of hydroxyproline (Hyp) in liver tissues; liver pathology and collagen staining; SOD activity and MDA, GSH, GSH-Px in liver tissues; protein expression of proliferating cell nuclear antigen (PCNA) in liver tissues. Serum ALT, AST, GGT, TBil significantly increased, and A1b decreased significantly in the model group. Hepatic Hyp significantly increased in the model group, whereas the index remarkably decreased in the Cordyceps polysaccharide group and the colchicine group. HE staining: the structure of normal hepatic lobules was damaged, with hepatocytes tumefaction and proliferation of connective tissues in portal tracts in the model group, while the Cordyceps polysaccharide group and the colchicine group recorded notable reduction in above pathological changes. Collagen staining: the model group showed hepatic lobule fibrous septum and many intact pseudolobules; while the Cordyceps polysaccharide group and the colchicine group witnessed decrease in collagen deposition. The model group showed significant decrease in SOD, GSH-Px and GSH and increase in MDA, whereas the Cordyceps polysaccharide group and the colchicine group recorded notable growth in GSH and GSH-Px. The model group showed significant decrease in protein expression of PCNA in liver tissues, while the Cordyceps polysaccharide group and the colchicine group showed significant reduction. Cordyceps polysaccharide can significantly inhibit DMN-induced liver fibrosis and lipid peroxidation in rats.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, lyophilized powder, ≥3,000 units/mg protein, Protein ≥95 % by biuret
Supelco
N-Nitrosodimethylamine (NDMA) solution, certified reference material, 200 μg/mL in methanol
γ-Glutamyl Transferase, ERM®, certified reference material
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, BioUltra, lyophilized powder, ≥4,500 units/mg protein, ≥97% (SDS-PAGE)
Sigma-Aldrich
Superoxide Dismutase bovine, recombinant, expressed in E. coli, lyophilized powder, ≥2500 units/mg protein, ≥90% (SDS-PAGE)
Sigma-Aldrich
Superoxide Dismutase from bovine liver, ammonium sulfate suspension, 2,000-6,000 units/mg protein (biuret)
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
trans-4-Hydroxy-L-proline, ≥99%
Sigma-Aldrich
γ-Glutamyltranspeptidase from porcine kidney, Type IV, 2.0-6.0 units/mg solid
Sigma-Aldrich
Glutamic-Pyruvic Transaminase from porcine heart, lyophilized powder, ≥75 units/mg protein
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioReagent, suitable for cell culture, ≥98.5%
Sigma-Aldrich
γ-Glutamyltranspeptidase from equine kidney, Type VI, 5-12 units/mg solid
Sigma-Aldrich
Glutamic-Pyruvic Transaminase from porcine heart, ammonium sulfate suspension, ≥75 units/mg protein
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, BioReagent, ≥3,000 units/mg protein, suitable for cell culture, lyophilized powder
Sigma-Aldrich
Glutamic-Oxalacetic Transaminase from porcine heart, Type I, ammonium sulfate suspension, 200-500 units/mg protein
Sigma-Aldrich
Superoxide Dismutase from bovine liver, lyophilized powder, ≥1500 units/mg protein
Supelco
trans-4-Hydroxy-L-proline, analytical standard
Sigma-Aldrich
Superoxide Dismutase from human erythrocytes, essentially salt-free, lyophilized powder, ≥2,500 units/mg protein
Sigma-Aldrich
Glutathione Peroxidase from bovine erythrocytes, lyophilized powder, ≥300 units/mg protein