Merck
Lipid phase behaviour under steady state conditions.
Faraday discussions (2013-06-29)
Christoffer Aberg, Emma Sparr, Håkan Wennerström
ABSTRAKT
At the interface between two regions, for example the air-liquid interface of a lipid solution, there can arise non-equilibrium situations. The water chemical potential corresponding to the ambient RH will, in general, not match the water chemical potential of the solution, and the gradients in chemical potential cause diffusional flows. If the bulk water chemical potential is close to a phase transition, there is the possibility of forming an interfacial phase with structures qualitatively different from those found in the bulk. Based on a previous analysis of this phenomenon in two component systems (C. Aberg, E. Sparr, K. J. Edler and H. Wennerström, Langmuir, 2009, 25, 12177), we here analyse the henomenon for three-component systems. The relevant transport equations are erived, and explicit results are given for some limiting cases. Then the formalism s applied conceptually to four different aqueous lipid systems, which in addition to water and a phospholipid contain (i) octyl glucoside, (ii) urea, (iii) heavy water, and (iv) sodium cholate as the third component. These four cases are chosen to illustrate (i) a method to use a micelle former to transport lipid to the interface where a multi-lamellar structure can form; (ii) to use a co-solvent to inhibit the formation of a gel phase at the interface; (iii) a method to form pure phospholipid multi-lamellar structures at the interface; (iv) a method to form a sequence of phases in the interfacial region. These four cases all have the character of theoretically based conjectures and it remains to investigate experimentally whether or not the conditions can be realized in practice.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Urea, European Pharmacopoeia (EP) Reference Standard