Merck
Comparison of the C14 urea breath test and histopathology in the diagnosis of Helicobacter pylori in the elderly.
JPMA. The Journal of the Pakistan Medical Association (2013-07-23)
Teslime Atli, Sevnaz Sahin, Bilge Ugur Arslan, Murat Varli, Ahmet Emre Yalcin, Sevgi Aras
ABSTRAKT
To compare the sensitivity and specificity of C[14] urea breath test with histopathological findings in biopsies obtained through endoscopy. In this cross-sectional study, conducted at the Medical Faculty Ankara University, Ankara, Turkey between January and October 2009, 100 patients aged 65 and above who fulfilled the criteria for inclusion were enrolled. Oesophago-gastroduodenoscopy was carried out on the patients after eight hours of fasting, using a Fujinon endoscope, by the same expert. C14 urea breath tests were carried out on all patients with a capsule containing 37 kBq(1micro Ci) C14 urea/citric acid. Evaluation of all data was carried out using SPSS 11.5. Categorical variables were compared through Pearson Chi squared or Fisher's exact test. P < 0.005 was taken as statistically significant. The average age of the patients was 71 +/- 5 years; 43 (43%) were male; and 57 (57%) were female. Helicobacter pylori positivity was found to be 35 (35%) in histopathological evaluation, and 36 (36%) in the C14 urea breath test. Of the 35 patients, 32 (91.4%) were found to be positive by the C14 urea breath test also. Of the 65 patients who were found to be negative on histopathology, 61 (93.8%) were also found to be Hp negative by the C14 urea breath test, and 4 (6.2%) were evaluated as false positive. A statistically significant correlation (p < 0.001) was found between histopathological and C14 urea breath test results. Compared with histopathology, the sensitivity of the urea breath test was 91.4%, and its specificity was 93.8%. A statistically significant (p < 0.001) relationship was found between histopathological results and the results of the C14 urea breath test. The sensitivity and specificity of the C[14] urea breath test in the group of elderly patients selected was high. The C14 urea breath test can be considered in the elderly group for diagnosis of Helicobacter pylori infection.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Urea, European Pharmacopoeia (EP) Reference Standard