Merck
A new approach for the characterization of proliferative cells in cestodes.
Experimental parasitology (2014-01-29)
M F Domínguez, U Koziol, V Porro, A Costábile, S Estrade, J Tort, M Bollati-Fogolin, E Castillo
ABSTRAKT
Cestodes show a remarkable proliferative capability that sustains the constant growth and differentiation of proglottids essential for their lifestyle. It is believed that a separate population of undifferentiated stem cells (the so-called germinative cells) are the only cells capable of proliferation during growth and development. The study of this particular cell subpopulation is hampered by the current lack of methods to isolate it. In this work, we developed a reproducible flow cytometry and cell sorting method to quantify and isolate the proliferating cells in the tetrathyridia larvae of the model cestode Mesocestoides corti, based on the DNA content of the cells. The isolated cells display the typical germinative cell morphology, and can be used for RNA isolation with a yield in the ng to μg range. We expect that this approach may facilitate the characterization of the germinative cells in M. corti and other model tapeworms.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trypsin from bovine pancreas, powder, ≥7,500 BAEE units/mg solid
Sigma-Aldrich
Trypsin from human pancreas, salt-free, lyophilized powder, vial of ≥1,000 BAEE units
Sigma-Aldrich
Trypsin from bovine pancreas, essentially salt-free, lyophilized powder, ≥9,000 BAEE units/mg protein, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, Type XI, lyophilized powder, ≥6,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, Type I, ~10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, ≥2,500 USP units/mg solid, meets USP testing specifications
Sigma-Aldrich
Trypsin Acetylated from bovine pancreas, Type V-S, ≥8,500 BAEE units/mg protein (biuret)
Sigma-Aldrich
Trypsin from porcine pancreas, ~1500 U/mg
Sigma-Aldrich
TrypZean® bovine, recombinant, expressed in corn, lyophilized powder, ≥3,350 units/mg solid (USP)
Sigma-Aldrich
Trypsin from porcine pancreas, tablet, 1 mg tablet
Sigma-Aldrich
Trypsin from bovine pancreas, suitable for protein sequencing, lyophilized powder
Sigma-Aldrich
Trypsin from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, Type II-S, 1,000-2,000 units/mg dry solid
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid