Merck
Epigallocatechin gallate counteracts oxidative stress in docosahexaenoxic acid-treated myocytes.
Biochimica et biophysica acta (2014-02-04)
Ester Casanova, Laura Baselga-Escudero, Aleix Ribas-Latre, Anna Arola-Arnal, Cinta Bladé, Lluís Arola, M Josepa Salvadó
ABSTRAKT
Skeletal muscle is a key organ of mammalian energy metabolism, and its mitochondria are multifunction organelles that are targets of dietary bioactive compounds. The goal of this work was to examine the regulation of mitochondrial dynamics, functionality and cell energy parameters using docosahexaenoic acid (DHA), epigallocatechin gallate (EGCG) and a combination of both in L6 myocytes. Compounds (at 25μM) were incubated for 4h. Cells cultured with DHA displayed less oxygen consumption with higher ADP/ATP ratio levels concomitant with downregulation of Cox and Ant1 gene expression. The disruption of energetic homeostasis by DHA, increases intracellular reactive oxygen species (ROS) levels and decreases mitochondrial membrane potential. The defence mechanism to counteract the excess of ROS production was by the upregulation of Ucp2, Ucp3 and MnSod gene expression. Moreover myocytes cultured with DHA had a higher mitochondrial mass with a higher proportion of large and elongated mitochondria, whereas the fission genes Drp1 and Fiss1 and the fusion gene Mfn2 were downregulated. In myocytes co-incubated with DHA and EGCG, ROS levels and the adenosine diphosphate (ADP)/adenosine triphosphate (ATP) ratio were similar to untreated myocytes and the decrease of oxygen consumption, higher mitochondrial mass and the overexpression of Ucp2 and Ucp3 genes were similar to the DHA-treated cells with also a higher amount of mitochondrial deoxyribonucleic acid (DNA), and reduced Drp1 and Fiss1 gene expression levels. In conclusion the addition of EGCG to DHA returned the cells to the control conditions in terms of mitochondrial morphology, energy and redox status, which were unbalanced in the DHA-treated myocytes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Alkohol odwodniony, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Supelco
(−)-Epicatechin, analytical standard
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (HPLC), powder
Epikatechina, primary reference standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
(+)-Catechin, analytical standard
Sigma-Aldrich
Fluo-3, suitable for fluorescence, ~70%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
cis-4,7,10,13,16,19-Docosahexaenoic acid, ≥98%
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Supelco
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
(−)-Epigallocatechin gallate, analytical standard
Galusan epigalokatechiny, primary reference standard
Sigma-Aldrich
Rhodamine 123, BioReagent, suitable for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
Rotenone, ≥95%
Supelco
Rotenone, PESTANAL®, analytical standard
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%