Merck
14C-urea breath test is safe for pediatric patients.
Nuclear medicine communications (2010-09-25)
Chander M Pathak, Balwinder Kaur, Krishan L Khanduja
ABSTRAKT
Helicobacter pylori (H. pylori) infection, quite prevalent in the developing countries, is considered to be one of the causative factors for various gastric pathologies and other nongastric diseases. It is believed that H. pylori infection is almost always acquired in early childhood and persists throughout life unless specific treatment is given. The (13/14)C-urea breath test (UBT) is now considered to be a 'gold standard' technique for the detection of H. pylori infection. However, because of the lack of facilities and high cost, the preferred nonradioactive ¹³C-UBT cannot be performed on pediatric patients in developing countries, whereas the radioactive ¹⁴C-UBT is not used on children because of the fear of radiation exposure. When using 37 kBq (1 μCi) of ¹⁴C-urea for the ¹⁴C-UBT, the patient is not exposed to more radiation than is acquired from the natural environment in one day, as almost all the ingested radioactivity is excreted from the body (urine and breath) within 72-120 h. This article reviews the importance of the ¹⁴C-UBT for the detection of H. pylori and justifies the radiation safety aspects of its use in children without any fear of 'radiation phobia' where the facility for ¹³C-UBT is lacking.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Urea, European Pharmacopoeia (EP) Reference Standard
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Supelco
Urea, analytical standard