Merck
  • Diagnostic performance of urea breath test, rapid urea test, and histology for Helicobacter pylori infection in patients with partial gastrectomy: a meta-analysis.
Diagnostic performance of urea breath test, rapid urea test, and histology for Helicobacter pylori infection in patients with partial gastrectomy: a meta-analysis.
Journal of clinical gastroenterology (2012-03-07)
Xue-Ying Tian, Hong Zhu, Jing Zhao, Qiang She, Guo-Xin Zhang
ABSTRAKT
Helicobacter pylori infection has been implicated in the pathogenesis of gastroduodenal diseases such as recurrent peptic ulceration and particularly residual stomach cancer in the postoperative stomach. To determine the performance of different commonly used tests for the diagnosis of H. pylori infection in patients after partial gastrectomy. A systematic literature search was conducted by searching the PubMed, EMBASE and ScienceDirect databases with relevant key words. Data extraction was independently performed by two reviewers. Meta-analyses were performed for the performance of the different tests including the sensitivities, specificities, likelihood ratios (LRs), diagnostic odds ratio diagnostic odds ratio, and the summary receiver operating characteristic summary receiver operating characteristic curve. The meta-analysis was performed by Meta-DiSc software. Studies showed a high degree of heterogeneity. Pooled sensitivity, specificity, LR+, LR- and diagnostic odds ratio for the different methods were: Urea breath test (9 studies): 0.77 (95% CI, 0.72-0.82), 0.89 (95% CI, 0.85-0.93), 6.32 (95% CI, 3.22-12.42), 0.27 (95% CI, 0.17-0.43), and 27.86 (95% CI, 13.27-58.49). Rapid urease test (7 studies): 0.79 (95% CI, 0.72-0.84), 0.94 (95% CI, 0.90-0.97), 10.21 (95% CI, 5.94-17.54), 0.28 (95% CI, 0.22-0.36) and 49.02 (95% CI, 24.24-99.14). Histology (3 studies): 0.93 (95% CI, 0.88-0.97), 0.85 (95% CI, 0.73-0.93), 5.88 (95% CI, 3.26-10.60), 0.09 (95% CI, 0.05- 0.15), and 97.28 (95% CI, 34.30-275.95). The corresponding summary receiver operating characteristic curves showed areas under the curves of 0.91, 0.93 and 0.96 and Q* values of 0.84, 0.86 and 0.91, respectively Among the three commonly used tests, histological examination performs the best, followed by the rapid urease test, for the diagnosis of H. pylori infection after partial gastrectomy. Thus, histology, preferably after the rapid urease test, is recommended for the diagnosis of H. pylori infection after partial gastrectomy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Urea, European Pharmacopoeia (EP) Reference Standard
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Supelco
Urea, analytical standard
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)