Merck
Prolonged ingestion of ovalbumin diet by sensitized mice improves the metabolic consequences induced by experimental food allergy.
Clinical and experimental immunology (2014-08-13)
N V Batista, R V S Pereira, M L M Noviello, L P A Dourado, D A Perez, G Foureaux, A J Ferreira, A V M Ferreira, D C Cara
ABSTRAKT
The prevalence of food allergy is rising in the western world. Allergen restriction is the chosen treatment in this condition, but continuous ingestion of the antigen has shown positive results in clinical trials. In a previous study, we have shown several allergic and metabolic alterations after 7 days of ovalbumin (OVA) ingestion by sensitized mice. The aim of this study was to investigate whether prolonged ingestion of antigen by sensitized mice would reverse the metabolic consequences caused by experimental food allergy. For this, allergic and metabolic parameters were analysed after prolonged ingestion of an OVA diet by OVA-sensitized mice. As shown previously, after 7 days of OVA consumption, sensitized mice showed increased serum levels of anti-OVA immunoglobulin (Ig)E and IgG1, aversion to the antigen ingestion, marked body and adipose tissue weight loss, followed by adipose tissue inflammation and decreased serum levels of adipokines, glucose and triglycerides. However, after 14 days of oral challenge, sensitized mice showed an anti-OVA IgE level similar to the mice that were only sensitized, but the specific IgG1 did not change. With this prolonged ingestion of OVA, sensitized mice were protected from OVA-induced anaphylaxis when the antigen was given systemically at a dose of 2 mg/animal. Moreover, various parameters analysed were significantly ameliorated, including adipose tissue inflammation, body and adipose tissue loss, as well as serum levels of adipokines and triglycerides. Therefore, our data suggest that prolonged ingestion of OVA by sensitized mice results in an improvement of the metabolic consequences caused by experimental food allergy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥90% (agarose gel electrophoresis)
Supelco
Albumin from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, powder, 62-88% (agarose gel electrophoresis)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
USP
Dekstroza, United States Pharmacopeia (USP) Reference Standard