Merck
Antioxidant vitamin intake and mortality: the Leisure World Cohort Study.
American journal of epidemiology (2015-01-01)
Annlia Paganini-Hill, Claudia H Kawas, María M Corrada
ABSTRAKT
To assess the relationship between antioxidant vitamin intake and all-cause mortality in older adults, we examined these associations using data from the Leisure World Cohort Study, a prospective study of residents of the Leisure World retirement community in Laguna Hills, California. In the early 1980s, participants (who were aged 44-101 years) completed a postal survey, which included details on use of vitamin supplements and dietary intake of foods containing vitamins A and C. Age-adjusted and multivariate-adjusted (for factors related to mortality in this cohort—smoking, alcohol intake, caffeine consumption, exercise, body mass index, and histories of hypertension, angina, heart attack, stroke, diabetes, rheumatoid arthritis, and cancer) hazard ratios for death were calculated using Cox regression for 8,640 women and 4,983 men (median age at entry, 74 years). During follow-up (1981-2013), 13,104 participants died (median age at death, 88 years). Neither dietary nor supplemental intake of vitamin A or vitamin C nor supplemental intake of vitamin E was significantly associated with mortality after multivariate adjustment. A compendium that summarizes previous findings of cohort studies evaluating vitamin intake and mortality is provided. Attenuation in the observed associations between mortality and antioxidant vitamin use after adjustment for confounders in our study and in previous studies suggests that such consumption identifies persons with other mortality-associated lifestyle and health risk factors.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sodium ascorbate, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Kwas askorbinowy, United States Pharmacopeia (USP) Reference Standard
Supelco
Retinol solution, 100 μg/mL ± 25% (Refer to COA) (Ethanol with 0.1% (w/v) BHT), ampule of 1 mL, reference material, Cerilliant®