Merck
  • Electro-acupuncture at Neiguan pretreatment alters genome-wide gene expressions and protects rat myocardium against ischemia-reperfusion.
Electro-acupuncture at Neiguan pretreatment alters genome-wide gene expressions and protects rat myocardium against ischemia-reperfusion.
Molecules (Basel, Switzerland) (2014-10-11)
Yan Huang, Sheng-Feng Lu, Chen-Jun Hu, Shu-Ping Fu, Wei-Xing Shen, Wan-Xin Liu, Qian Li, Ning Wang, Su-Yun He, Fan-Rong Liang, Bing-Mei Zhu
ABSTRAKT
This study investigated genome-wide gene expressions and the cardioprotective effects of electro-acupuncture pretreatment at the PC6 Neiguan acupoint on myocardial ischemia reperfusion (I/R) injury. Male SD rats were randomly divided into four groups: sham operation (SO), I/R, electro-acupuncture at the PC6 Neiguan acupoint pretreatment (EA) and electro-acupuncture at non-acupoint pretreatment (NA). Compared with the I/R group, the survival rate of the EA group was significantly increased, the arrhythmia score, infarction area, serum concentrations of CK, LDH and CK-Mb and plasma level of cTnT were significantly decreased. RNA-seq results showed that 725 genes were up-regulated and 861 genes were down-regulated under I/R conditions compared to the SO group; both EA and NA reversed some of these gene expression levels (592 in EA and 238 in NA group). KEGG pathway analysis indicated that these genes were involved in multiple pathways, including ECM, MAPK signaling, apoptosis, cytokine and leukocyte pathways. In addition, some pathways were uniquely regulated by EA, but not NA pretreatment, such as oxidative stress, cardiac muscle contraction, gap junction, vascular smooth muscle contraction, hypertrophic, NOD-like receptor, and P53 and B-cell receptor pathways. This study was first to reveal the gene expression signatures of acute myocardial I/R injury and electro-acupuncture pretreatment in rats.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, Molecular Biology, suitable for electrophoresis, ≥99.5%
Supelco
Acrylamide solution, 40% in H2O
Sigma-Aldrich
N,N′-Methylenebisacrylamide solution, suitable for electrophoresis, 2% in H2O
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered