Merck
Validation of methylation markers for diagnosis of oral cavity cancer.
European journal of cancer (Oxford, England : 1990) (2015-02-18)
L M R B Arantes, A C de Carvalho, M E Melendez, C C Centrone, J F Góis-Filho, T N Toporcov, D N Caly, E H Tajara, E M Goloni-Bertollo, A L Carvalho
ABSTRAKT
Activation of proto-oncogenes and inactivation of tumour suppressor genes are the major genetic alterations involved in carcinogenesis. The increase in methylation at the promoter region of a tumour suppressor gene can lead to gene inactivation, selecting cells with proliferative advantage. Thus, promoter hypermethylation is considered a marker in a variety of malignant tumours, including oral cavity. The methylation pattern of eight genes was evaluated in 40 oral cavity squamous cell carcinomas (OSCCs) and 40 saliva samples from healthy individuals by Q-MSP. Different combinations of genes were also assessed in order to identify gene panels that could better distinguish between OSCC and saliva samples. CCNA1, DAPK, DCC and TIMP3 methylation were highly specific for being found in the OSCC samples. Moreover, the combination of these genes improved detection when compared with single markers, reaching values of 92.5% for sensitivity and specificity (when using the panel CCNA1, DCC, TIMP3). Moreover, DAPK, DCC and TIMP3 were hypermethylated in nearly 90% of clinically T1 and T2 cases. The pursuing of this panel of hypermethylated genes is an important tool for the detection of individuals with OSCC. Moreover, the identification of these markers in early stages of OSCC shows the feasibility of using the panel on saliva as possible biomarkers for early diagnosis. The lack of association between the methylation status of these genes and clinical characteristics shows that they are able to distinguish OSCC cases irrespective of social and clinical factors (gender, age, human papillomavirus (HPV) status, clinical stage, vascular embolisation and perineural invasion).
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Supelco
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
tert-Butyl acetoacetate, reagent grade, 95%
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Ammonium sulfate, Molecular Biology, ≥99.0%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)