Merck
MspA-Mycobacterium tuberculosis-transformant with reduced virulence: the "unbirthday paradigm".
Microbial pathogenesis (2014-09-10)
Otmane Lamrabet, Eric Ghigo, Jean-Louis Mège, Hubert Lepidi, Claude Nappez, Didier Raoult, Michel Drancourt
ABSTRAKT
Expressing mspA porin gene from Mycobacterium smegmatis in Mycobacterium tuberculosis attenuated this pathogen. Intracellular growth of the transformants into free-living amoeba and murine and human macrophages decreased. Furthermore, transformants decreased the microbicidal program of human monocyte-derived macrophages. BALB/c mice inoculated with transformants exhibited higher weights, lower histological lesions and lower M. tuberculosis inoculum in the liver, spleen and lungs than control mice challenged with wild-type M. tuberculosis. Preliminary evaluation indicated that mice inoculated with this transformant showed higher weights and lower numbers of lung nodules and tissular mycobacteria than control mice when challenged with wild-type M. tuberculosis. Similar to the paradoxical "unbirthday" gift coined by Lewis Carroll in Alice's Adventures in Wonderland, adding mspA gene reduced the virulence of M. tuberculosis and yielded a protective effect. Lost of non-virulence genes is a mechanism for virulence in mycobacteria. Engineering non-virulence genes in M. tuberculosis may yield strains with decreased virulence and increased immunogenicity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Gliceryna, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, FCC, FG
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®