Merck
In vivo selective imaging and inhibition of leukemia stem-like cells using the fluorescent carbocyanine derivative, DiOC5(3).
Biomaterials (2015-03-31)
Beibei Zhang, Yasuhito Shimada, Junya Kuroyanagi, Michiko Ariyoshi, Tsuyoshi Nomoto, Taichi Shintou, Noriko Umemoto, Yuhei Nishimura, Takeshi Miyazaki, Toshio Tanaka
ABSTRAKT
Elimination of leukemia stem cells (LSCs) is necessary for the destruction of malignant cell populations. Owing to the very small number of LSCs in leukemia cells, xenotransplantation studies are difficult in terms of functionally and pathophysiologically replicating clinical conditions of cell culture experiments. There is currently a limited number of lead compounds that target LSCs. Using the LSC-xenograft zebrafish screening method we previously developed, we found that the fluorescent compound 3,3'-dipentyloxacarbocyanine iodide (DiOC5(3)) selectively marked LSCs and suppressed their proliferation in vivo and in vitro. DiOC5(3) had no obvious toxicity to human umbilical cord blood CD34+ progenitor cells and normal zebrafish. It accumulated in mitochondria through organic anion transporter polypeptides that are overexpressed in the plasma membrane of LSCs, and induced apoptosis via ROS overproduction. DiOC5(3) also inhibited the nuclear translocation of NF-κB through the downregulation of LSC-selective pathways, as indicated from DNA microarray analysis. In summary, DiOC5(3) is a new type of anti-LSC compound available for diagnostic imaging and therapeutics that has the advantage of being a single fluorescent chemical.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
2-Phenoxyethanol, ≥99%
Supelco
2-Phenoxyethanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
2-Phenoxyethanol, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Ethylene glycol monophenyl ether, ≥90%
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, Molecular Biology
Sigma-Aldrich
Magnesium sulfate solution, Molecular Biology, 1.00 M±0.04 M