Merck
Leishmanial lipid suppresses the bacterial endotoxin-induced inflammatory response with attenuation of tissue injury in sepsis.
Journal of leukocyte biology (2014-04-04)
Nabanita Chatterjee, Subhadip Das, Dipayan Bose, Somenath Banerjee, Tarun Jha, Krishna Das Saha
ABSTRAKT
The use of live, attenuated, or genetically modified microbes or their cellular component(s) or metabolites has begun to emerge as a potential new approach in medicinal research to deliver biologically active entities. Thus, advancing our knowledge of such microbe-mediated therapy may suggest new avenues for therapeutic intervention in many diseases. We had earlier reported that the total lipid of attenuated Leishmania donovani suppressed the inflammatory responses in rheumatoid arthritis patients. Our present study reveals that the pLLD, isolated from pathogenic L. donovani, decreases the inflammatory level of bacterial endotoxin in stimulated mouse macrophages, as also in the in vivo murine system. It exerts the activity by reducing the level of different mediators, such as cytokine-chemokine(s). It also suppresses the expression of the ubiquitous transcription factor NF-κBp65 in stimulated macrophage cells, improves the endotoxin-associated liver damage, reduces the vascular permeability factors, such as VEGF, and suppresses the expression of cell adhesion molecules, including ICAM-1, VCAM-1, PECAM-1, P-selectin, and E-selectin, in liver of septic mice. These findings indicate that pLLD may prove to be a potential anti-inflammatory agent and protect from endotoxin-induced sepsis in hepatic impairment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Bromek cetrymonium, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)