Merck
Programmable transdermal clonidine delivery through voltage-gated carbon nanotube membranes.
Journal of pharmaceutical sciences (2014-05-03)
Caroline Strasinger, Kalpana S Paudel, Ji Wu, Dana Hammell, Raghotham R Pinninti, Bruce J Hinds, Audra Stinchcomb
ABSTRAKT
Oral dosage forms and traditional transdermal patches are inadequate for complex clonidine therapy dosing schemes, because of the variable dose/flux requirement for the treatment of opioid withdrawal symptoms. The purpose of this study was to evaluate the in vitro transdermal flux changes of clonidine in response to alterations in carbon nanotube (CNT) delivery rates by applying various electrical bias. Additional skin diffusion studies were carried out to demonstrate the therapeutic feasibility of the system. This study demonstrated that application of a small electrical bias (-600 mV) to the CNT membrane on the skin resulted in a 4.7-fold increase in clonidine flux as compared with no bias (0 mV) application. The high and low clonidine flux values were very close to the desired variable flux of clonidine for the treatment of opioid withdrawal symptoms. Therapeutic feasibility studies demonstrated that CNT membrane served as the rate-limiting step to clonidine diffusion and lag and transition times were suitable for the clonidine therapy. Skin elimination studies revealed that clonidine depletion from the skin would not negatively affect clonidine therapy. Overall, this study showed that clonidine administration difficulties associated with the treatment of opiate withdrawal symptoms can be reduced with the programmable CNT membrane transdermal system.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)