Merck
Prevalence of chlorhexidine-resistant methicillin-resistant Staphylococcus aureus following prolonged exposure.
Antimicrobial agents and chemotherapy (2014-05-21)
Carey D Schlett, Eugene V Millar, Katrina B Crawford, Tianyuan Cui, Jeffrey B Lanier, David R Tribble, Michael W Ellis
ABSTRAKT
Chlorhexidine has been increasingly utilized in outpatient settings to control methicillin-resistant Staphylococcus aureus (MRSA) outbreaks and as a component of programs for MRSA decolonization and prevention of skin and soft-tissue infections (SSTIs). The objective of this study was to determine the prevalence of chlorhexidine resistance in clinical and colonizing MRSA isolates obtained in the context of a community-based cluster-randomized controlled trial for SSTI prevention, during which 10,030 soldiers were issued chlorhexidine for body washing. We obtained epidemiological data on study participants and performed molecular analysis of MRSA isolates, including PCR assays for determinants of chlorhexidine resistance and high-level mupirocin resistance and pulsed-field gel electrophoresis (PFGE). During the study period, May 2010 to January 2012, we identified 720 MRSA isolates, of which 615 (85.4%) were available for molecular analysis, i.e., 341 clinical and 274 colonizing isolates. Overall, only 10 (1.6%) of 615 isolates were chlorhexidine resistant, including three from the chlorhexidine group and seven from nonchlorhexidine groups (P > 0.99). Five (1.5%) of the 341 clinical isolates and five (1.8%) of the 274 colonizing isolates harbored chlorhexidine resistance genes, and four (40%) of the 10 possessed genetic determinants for mupirocin resistance. All chlorhexidine-resistant isolates were USA300. The overall prevalence of chlorhexidine resistance in MRSA isolates obtained from our study participants was low. We found no association between extended chlorhexidine use and the prevalence of chlorhexidine-resistant MRSA isolates; however, continued surveillance is warranted, as this agent continues to be utilized for infection control and prevention efforts.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Mannitol, European Pharmacopoeia (EP) Reference Standard
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.