Merck
Rapid and reproducible analysis of thiocyanate in real human serum and saliva using a droplet SERS-microfluidic chip.
Biosensors & bioelectronics (2014-06-29)
Lei Wu, Zhuyuan Wang, Shenfei Zong, Yiping Cui
ABSTRAKT
As thiocyanate (SCN(-)) acts as an important biomarker in human health assessment, there remains an urgent need to realize rapid and reproducible analysis of SCN(-) in body fluids. Here, a droplet microfluidic device has been designed and fabricated for SCN(-) detection in real human serum and saliva using the surface enhanced Raman scattering (SERS) technique. Only a few minutes are needed for the whole detection process which simply cost a few microliters of real sample. Gold@silver core-shell nanorods (Au@Ag NRs) with a large SERS enhancement factor were selected to capture SCN(-) ions in body fluids. The intensity of SERS peak at around 2100 cm(-1), which originates from the -C≡N stretching mode, was used to indicate the concentrations of SCN(-) ions. Importantly, by generating a droplet environment for mixing reagents and acquiring signals, this microfluidic platform possesses the advantages of an improved reproducibility and reduced time consumption. For practical applications, the SERS-microfluidic system is capable to achieve rapid analysis of SCN(-) in the presence of human serum, which is very important for realizing the detection in real biological samples. Additionally, SCN(-) in saliva samples was detected in the SERS-microfluidic chip and the results provide useful information for distinguishing between smokers and nonsmokers.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Bromek cetrymonium, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Potassium thiocyanate, BioXtra, ≥99.0%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Potassium thiocyanate, ReagentPlus®, ≥99.0%
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
VenPure® SF, powder