Merck
  • Population genetics of Trypanosoma brucei gambiense in sleeping sickness patients with treatment failures in the focus of Mbuji-Mayi, Democratic Republic of the Congo.
Population genetics of Trypanosoma brucei gambiense in sleeping sickness patients with treatment failures in the focus of Mbuji-Mayi, Democratic Republic of the Congo.
Infection, genetics and evolution : journal of molecular epidemiology and evolutionary genetics in infectious diseases (2014-12-24)
Patient Pati Pyana, Modou Sere, Jacques Kaboré, Thierry De Meeûs, Annette MacLeod, Bruno Bucheton, Nick Van Reet, Philippe Büscher, Adrien Marie Gaston Belem, Vincent Jamonneau
ABSTRAKT
Human African trypanosomiasis (HAT) in the Democratic Republic of the Congo (DRC) is caused by the protozoan Trypanosoma brucei gambiense. Until recently, all patients in the second or neurological stage of the disease were treated with melarsoprol. At the end of the past and the beginning of the present century, alarmingly high relapse rates in patients treated with melarsoprol were reported in isolated HAT foci. In the Mbuji-Mayi focus of DRC, a particular mutation that confers cross resistance for pentamidine and melarsoprol was recently found for all strains studied. Nevertheless, treatment successfully cured a significant proportion of patients. To check for the existence of other possible genetic factors of the parasites, we genotyped trypanosomes isolated from patients before and after treatment (relapsing patients) with eight microsatellite markers. We found no evidence of any genetic correlation between parasite genotype and treatment outcome and we concluded that relapse or cure probably depend more on patients' factors such as disease progression, nutritional or immunological status or co-infections with other pathogens. The existence of a melarsoprol and pentamidine resistance associated mutation at such high rates highlights an increasing problem, even for other drugs, especially those using the same transporters as melarsoprol and pentamidine.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
2-Mercaptoethanol, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
5-Carboxyfluorescein, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Hexarelin, ≥90% (HPLC)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
2-(Diethylamino)ethanol, ≥99.5%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Cyclophosphamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)