Merck
Synthesis and antitumor activity of novel per-butyrylated glycosides of podophyllotoxin and its derivatives.
Bioorganic & medicinal chemistry (2015-03-07)
Cheng-Ting Zi, Dan Yang, Fa-Wu Dong, Gen-Tao Li, Yan Li, Zhong-Tao Ding, Jun Zhou, Zi-Hua Jiang, Jiang-Miao Hu
ABSTRAKT
A series of perbutyrylated glycosides of podophyllotoxin and its derivatives were synthesized and evaluated for their antitumor activity in vitro. Most of them exhibit cytotoxic activity against a panel of five human cancer cell lines (HL-60, SMMC-7721, A-549, MCF-7, SW480) using MTT assays. Among the synthesized compounds, epipodophyllotoxin α-d-galactopyranoside 8b, epipodophyllotoxin α-d-arabinopyranoside 8e, and podophyllotoxin β-d-glucopyranoside 11a show the highest potency of anticancer activity with their IC50 values ranging from 0.14 to 1.69μM. Structure activity relationship analysis indicates that the type of glycosidic linkage, the configuration at C-4 of the podophyllotoxin scaffold, and the substitution at 4'-position (OH vs OCH3) can all have significant effect on the potency of their anticancer activity. Several compounds are more active than the control drugs Etoposide and Cisplatin, suggesting their potential as anticancer agents for further development.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Sigma-Aldrich
Maltose solution, BioReagent, ~20% in H2O, Molecular Biology
Sigma-Aldrich
Podophyllotoxin
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dekstroza, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99%
Millipore
D-(+)-Mannose, ≥99.0% (sum of enantiomers, HPLC), suitable for microbiology
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-(+)-Glucose, analytical standard