Merck
  • Infectious agents and inflammation in donated hearts and dilated cardiomyopathies related to cardiovascular diseases, Chagas' heart disease, primary and secondary dilated cardiomyopathies.
Infectious agents and inflammation in donated hearts and dilated cardiomyopathies related to cardiovascular diseases, Chagas' heart disease, primary and secondary dilated cardiomyopathies.
International journal of cardiology (2014-12-03)
Sandrigo Mangini, Maria de Lourdes Higuchi, Joyce Tiyeko Kawakami, Marcia Martins Reis, Renata Nishiyama Ikegami, Suely Aparecida Pinheiro Palomino, Pablo Maria Alberto Pomerantzeff, Alfredo Inácio Fiorelli, Fabiana Goulart Marcondes-Braga, Fernando Bacal, Sílvia Moreira Ayub Ferreira, Victor Sarli Issa, Germano Emílio Conceição Souza, Paulo Roberto Chizzola, Edimar Alcides Bocchi
ABSTRAKT
Clinical and experimental conflicting data have questioned the relationship between infectious agents, inflammation and dilated cardiomyopathy (DCM). The aim of this study was to determine the frequency of infectious agents and inflammation in endomyocardial biopsy (EMB) specimens from patients with idiopathic DCM, explanted hearts from different etiologies, including Chagas' disease, compared to donated hearts. From 2008 to 2011, myocardial samples from 29 heart donors and 55 patients with DCMs from different etiologies were studied (32 idiopathic, 9 chagasic, 6 ischemic and 8 other specific etiologies). Inflammation was investigated by immunohistochemistry and infectious agents by immunohistochemistry, molecular biology, in situ hybridization and electron microscopy. There were no differences regarding the presence of macrophages, expression of HLA class II and ICAM-I in donors and DCM. Inflammation in Chagas' disease was predominant. By immunohistochemistry, in donors, there was a higher expression of antigens of enterovirus and Borrelia, hepatitis B and C in DCMs. By molecular biology, in all groups, the positivity was elevated to microorganisms, including co-infections, with a higher positivity to adenovirus and HHV6 in donors towards DCMs. This study was the first to demonstrate the presence of virus in the heart tissue of chagasic DCM. The presence of inflammation and infectious agents is frequent in donated hearts, in the myocardium of patients with idiopathic DCM, myocardial dysfunction related to cardiovascular diseases, and primary and secondary cardiomyopathies, including Chagas' disease. The role of co-infection in Chagas' heart disease physiopathology deserves to be investigated in future studies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium trichloroacetate, 97%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Informacje o cenach i dostępności nie są obecnie dostępne.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Aurintricarboxylic acid, practical grade, ≥85% (titration), powder
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Informacje o cenach i dostępności nie są obecnie dostępne.
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Informacje o cenach i dostępności nie są obecnie dostępne.