Merck
Rapid identification and quantitative validation of a caffeine-degrading pathway in Pseudomonas sp. CES.
Journal of proteome research (2014-10-29)
Chi Li Yu, Ryan M Summers, Yalan Li, Sujit Kumar Mohanty, Mani Subramanian, R Marshall Pope
ABSTRAKT
Understanding the genes and enzymes involved in caffeine metabolism can lead to applications such as production of methylxanthines and environmental waste remediation. Pseudomonas sp. CES may provide insights into these applications, since this bacterium degrades caffeine and thrives in concentrations of caffeine that are three times higher (9.0 g L(-1)) than the maximum tolerable levels of other reported bacteria. We took a novel approach toward identifying the enzymatic pathways in Pseudomonas sp. CES that metabolize caffeine, which largely circumvented the need for exhaustive isolation of enzymes and the stepwise reconstitution of their activities. Here we describe an optimized, rapid alternative strategy based on multiplexed LC-MS/MS assays and show its application by discovering caffeine-degrading enzymes in the CES strain based on quantitative comparison of proteomes from bacteria grown in the absence and presence of caffeine, the latter condition of which was found to have a highly induced capacity for caffeine degradation. Comparisons were made using stable isotope dimethyl labeling, differences in the abundance of particular proteins were substantiated by reciprocal labeling experiments, and the role of the identified proteins in caffeine degradation was independently verified by genetic sequencing. Overall, multiple new components of a N-demethylase system were identified that resulted in rapid pathway validation and gene isolation using this new approach.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
SAFC
BIS-TRIS
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioUltra, Molecular Biology, ≥99.5% (titration)
SAFC
MOPS
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O