Merck
Astrocytic dynamin-like protein 1 regulates neuronal protection against excitotoxicity in Parkinson disease.
The American journal of pathology (2014-12-09)
Jake G Hoekstra, Travis J Cook, Tessandra Stewart, Hayley Mattison, Max T Dreisbach, Zachary S Hoffer, Jing Zhang
ABSTRAKT
Mitochondrial dynamics has recently become an area of piqued interest in neurodegenerative disorders, including Parkinson disease (PD); however, the contribution of astrocytes to these disorders remains unclear. Here, we show that the level of dynamin-like protein 1 (Dlp1; official name DNM1L), which promotes mitochondrial fission, is lower in astrocytes from the brains of PD patients, and that decreased astrocytic Dlp1 likely represents a relatively early event in PD pathogenesis. In support of this conclusion, we show that Dlp1 knockdown dramatically affects mitochondrial morphological characteristics and localization in astrocytes, impairs the ability of astrocytes to adequately protect neurons from the excitotoxic effects of glutamate, and increases intracellular Ca(2+) in response to extracellular glutamate, resulting from compromised intracellular Ca(2+) buffering. Taken together, our results suggest that astrocytic mitochondrial Dlp1 is a key protein in mitochondrial dynamics and decreased Dlp1 may interfere with neuron survival in PD by disrupting Ca(2+)-coupled glutamate uptake.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
USP
Alkohol odwodniony, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
DL-Tyrosine, 99%
SAFC
L-Cysteine
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
L-Glutamic acid monosodium salt hydrate, ≥99% (HPLC), powder
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Tyrosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
L-Glutamic acid monosodium salt hydrate, analytical standard
Sigma-Aldrich
L-Glutamic acid monosodium salt hydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, Wacker Chemie AG, ≥98.0%
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma