Merck
Biopharmaceutical characterization of nebulized antimicrobial agents in rats: 1. Ciprofloxacin, moxifloxacin, and grepafloxacin.
Antimicrobial agents and chemotherapy (2014-05-07)
Aline Vidal Lacerda Gontijo, Julien Brillault, Nicolas Grégoire, Isabelle Lamarche, Patrice Gobin, William Couet, Sandrine Marchand
ABSTRAKT
The aim of this study was to evaluate the biopharmaceutical characteristics of three fluoroquinolones (FQs), ciprofloxacin (CIP), moxifloxacin (MXF), and grepafloxacin (GRX), after delivery via a nebulized aerosol to rats. Bronchoalveolar lavages (BAL) were conducted 0.5, 2, 4, and 6 h after FQ intravenous administration and nebulized aerosol delivery to estimate epithelial lining fluid (ELF) drug concentrations. Plasma drug concentrations were also measured, and profiles of drug concentrations versus time after intravenous administration and nebulized aerosol delivery were virtually superimposable, attesting for rapid and complete systemic absorption of FQs. ELF drug concentrations were systematically higher than corresponding plasma drug concentrations, whatever the route of administration, and average ELF-to-unbound plasma drug concentration ratios post-distribution equilibrium did not change significantly between the ways of administration and were equal: 4.0 ± 5.3 for CIP, 12.6 ± 7.3 for MXF, and 19.1 ± 10.5 for GRX (means ± standard deviations). The impact of macrophage lysis on estimated ELF drug concentrations was significant for GRX but reduced for MXF and CIP; therefore, simultaneous pharmacokinetic modeling of plasma and ELF drug concentrations was only performed for the latter two drugs. The model was characterized by a fixed volume of ELF (VELF), passive diffusion clearance (QELF), and active efflux clearance (CLout) between plasma and ELF, indicating active efflux transport systems. In conclusion, this study demonstrates that ELF drug concentrations of these three FQs are several times higher than plasma drug concentrations, probably due to the presence of efflux transporters at the pulmonary barrier level, but no biopharmaceutical advantage of FQ nebulization was observed compared with intravenous administration.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Moxifloxacin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Moxifloxacin hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Urea, European Pharmacopoeia (EP) Reference Standard
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Supelco
Urea, analytical standard
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)