Merck
Extraocular muscle satellite cells are high performance myo-engines retaining efficient regenerative capacity in dystrophin deficiency.
Developmental biology (2014-09-23)
Pascal Stuelsatz, Andrew Shearer, Yunfei Li, Lindsey A Muir, Nicholas Ieronimakis, Qingwu W Shen, Irina Kirillova, Zipora Yablonka-Reuveni
ABSTRAKT
Extraocular muscles (EOMs) are highly specialized skeletal muscles that originate from the head mesoderm and control eye movements. EOMs are uniquely spared in Duchenne muscular dystrophy and animal models of dystrophin deficiency. Specific traits of myogenic progenitors may be determinants of this preferential sparing, but very little is known about the myogenic cells in this muscle group. While satellite cells (SCs) have long been recognized as the main source of myogenic cells in adult muscle, most of the knowledge about these cells comes from the prototypic limb muscles. In this study, we show that EOMs, regardless of their distinctive Pax3-negative lineage origin, harbor SCs that share a common signature (Pax7(+), Ki67(-), Nestin-GFP(+), Myf5(nLacZ+), MyoD-positive lineage origin) with their limb and diaphragm somite-derived counterparts, but are remarkably endowed with a high proliferative potential as revealed in cell culture assays. Specifically, we demonstrate that in adult as well as in aging mice, EOM SCs possess a superior expansion capacity, contributing significantly more proliferating, differentiating and renewal progeny than their limb and diaphragm counterparts. These robust growth and renewal properties are maintained by EOM SCs isolated from dystrophin-null (mdx) mice, while SCs from muscles affected by dystrophin deficiency (i.e., limb and diaphragm) expand poorly in vitro. EOM SCs also retain higher performance in cell transplantation assays in which donor cells were engrafted into host mdx limb muscle. Collectively, our study provides a comprehensive picture of EOM myogenic progenitors, showing that while these cells share common hallmarks with the prototypic SCs in somite-derived muscles, they distinctively feature robust growth and renewal capacities that warrant the title of high performance myo-engines and promote consideration of their properties for developing new approaches in cell-based therapy to combat skeletal muscle wasting.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Aphidicolin, analytical standard
Supelco
Pyridoxine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Pyridoxine Hydrochloride (B6), analytical standard
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Eosin Y disodium salt, certified by the BSC
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Eosin Y disodium salt, ≥85 % by HPLC
Sigma-Aldrich
Pyridoxine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Pyridoxine hydrochloride, meets USP testing specifications
Sigma-Aldrich
Pyridoxine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Eosin Y, 75% (HPLC)
Sigma-Aldrich
Eosin Y solution, 5 wt. % in H2O
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Methylbutane, suitable for HPLC, ≥99.5%
Pyridoxine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
2-Methylbutane, anhydrous, ≥99%
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Supelco
2-Methylbutane, analytical standard
Sigma-Aldrich
Anti-Laminin Antibody, Chemicon®, from rabbit
USP
Pyridoxine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%