Merck
A spontaneous animal model of intestinal dysmotility evoked by inflammatory nitrergic dysfunction.
PloS one (2014-05-14)
Tatsuhiro Masaoka, Tim Vanuytsel, Christophe Vanormelingen, Sebastien Kindt, Shadea Salim Rasoel, Werend Boesmans, Gert De Hertogh, Ricard Farré, Pieter Vanden Berghe, Jan Tack
ABSTRAKT
Recent reports indicate the presence of low grade inflammation in functional gastrointestinal disorders (FGID), in these cases often called "post-inflammatory" FGIDs. However, suitable animal models to study these disorders are not available. The Biobreeding (BB) rat consists of a diabetes-resistant (BBDR) and a diabetes-prone (BBDP) strain. In the diabetes-prone strain, 40-60% of the animals develop diabetes and concomitant nitrergic dysfunction. Our aim was to investigate the occurrence of intestinal inflammation, nitrergic dysfunction and intestinal dysmotility in non-diabetic animals. Jejunal inflammation (MPO assay, Hematoxylin&Eosin staining and inducible nitric oxide synthase (iNOS) mRNA expression), in vitro jejunal motility (video analysis) and myenteric neuronal numbers (immunohistochemistry) were assessed in control, normoglycaemic BBDP and diabetic BBDP rats. To study the impact of iNOS inhibition on these parameters, normoglycaemic BBDP rats were treated with aminoguanidine. Compared to control, significant polymorphonuclear (PMN) cell infiltration, enhanced MPO activity, increased iNOS mRNA expression and a decreased ratio of nNOS to Hu-C/D positive neurons were observed in both normoglycaemic and diabetic BBDP rats. Aminoguanidine treatment decreased PMN infiltration, iNOS mRNA expression and MPO activity. Moreover, it restored the ratio of nNOS to Hu-C/D positive nerves in the myenteric plexus and decreased the abnormal jejunal elongation and dilation observed in normoglycaemic BBDP rats. Aminoguanidine treatment counteracts the inflammation-induced nitrergic dysfunction and prevents dysmotility, both of which are independent of hyperglycaemia in BB rats. Nitrergic dysfunction may contribute to the pathophysiology of "low-grade inflammatory" FGIDs. Normoglycaemic BBDP rats may be considered a suitable animal model to study the pathogenesis of FGIDs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Tranexamic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
trans-4-(Aminomethyl)cyclohexanecarboxylic acid, 97%
USP
Kwas traneksamowy, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Toluidine Blue, suitable for microscopy, 8.74% (ZN (THEORY))
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet