Merck
Assessment of in vitro cyto/genotoxicity of sequentially treated electroplating effluent on the human hepatocarcinoma HuH-7 cell line.
Mutation research. Genetic toxicology and environmental mutagenesis (2014-02-15)
Umesh Chandra Naik, Mihir Tanay Das, Swati Sauran, Indu Shekhar Thakur
ABSTRAKT
The present study compares in vitro toxicity of electroplating effluent after the batch treatment process with that obtained after the sequential treatment process. Activated charcoal prepared from sugarcane bagasse through chemical carbonization, and tolerant indigenous bacteria, Bacillus sp. strain IST105, were used individually and sequentially for the treatment of electroplating effluent. The sequential treatment involving activated charcoal followed by bacterial treatment removed 99% of Cr(VI) compared with the batch processes, which removed 40% (charcoal) and 75% (bacteria), respectively. Post-treatment in vitro cyto/genotoxicity was evaluated by the MTT test and the comet assay in human HuH-7 hepatocarcinoma cells. The sequentially treated sample showed an increase in LC50 value with a 6-fold decrease in comet-assay DNA migration compared with that of untreated samples. A significant decrease in DNA migration and an increase in LC50 value of treated effluent proved the higher effectiveness of the sequential treatment process over the individual batch processes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, Molecular Biology
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Potassium dichromate, 99.98% trace metals basis
Supelco
Perchloric acid, 0.01 M HClO4 in water (0.01N), eluent for IC