Merck
Suppression of adipocyte hypertrophy by polymethoxyflavonoids isolated from Kaempferia parviflora.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2014-03-19)
Yui Okabe, Tsutomu Shimada, Takumi Horikawa, Kaoru Kinoshita, Kiyotaka Koyama, Koji Ichinose, Masaki Aburada, Kunio Takahashi
ABSTRAKT
We previously demonstrated that ethyl acetate extracts of Kaempferia parviflora Wall. Ex Baker (KPE) improve insulin resistance in TSOD mice and showed that its components induce differentiation and adipogenesis in 3T3-L1 preadipocytes. The present study was undertaken to examine whether KPE and its isolated twelve components suppress further lipid accumulation in 3T3-L1 mature adipocytes. KPE reduced intracellular triglycerides in mature adipocytes, as did two of its components, 3,5,7,3',4'-pentamethoxyflavone and 5,7,4'-trimethoxyflavone. Shrinkage of lipid droplets in mature adipocytes was observed, and mRNA expression levels of adipose tissue triglyceride lipase (ATGL) and hormone-sensitive lipase (HSL) were up-regulated by these two polymethoxyflavonoids (PMFs). Furthermore, the protein expression level of ATGL and the release level of glycerol into the cell culture medium increased. In contrast, the peroxisome proliferator-activated receptor γ (PPARγ) agonist, troglitazone, did not decrease intracellular triglycerides in mature adipocytes, and the mRNA expression level of PPARγ was not up-regulated in mature adipocytes treated with the two active PMFs. Therefore, suppression of lipid accumulation in mature adipocytes is unlikely to be enhanced by transcriptional activation of PPARγ. These results suggest that KPE and its active components enhance lipolysis in mature adipocytes by activation of ATGL and HSL independent of PPARγ transcription, thus preventing adipocyte hypertrophy. On the other hand, the full hydroxylated flavonoid quercetin did not show the suppressive effects of lipid accumulation in mature adipocyte in the same conditions. Consequently, methoxy groups in the flavones are important for the activity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Gliceryna, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Nobiletin, ≥97%
Sigma-Aldrich
Troglitazone, ≥98% (HPLC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, FCC, FG