Merck
Binding kinetics, potency, and selectivity of the hepatitis C virus NS3 protease inhibitors GS-9256 and vedroprevir.
Biochimica et biophysica acta (2014-08-21)
Ona Barauskas, Amoreena C Corsa, Ruth Wang, Scott Hluhanich, Debi Jin, Magdeleine Hung, Huiling Yang, William E Delaney, Brian E Schultz
ABSTRAKT
GS-9256 and vedroprevir are inhibitors of the hepatitis C virus NS3 protease enzyme, an important drug target. The potency, selectivity, and binding kinetics of the two compounds were determined using in vitro biochemical assays. Potency of the compounds against NS3 protease and selectivity against a panel of mammalian proteases were determined through steady-state enzyme kinetics. Binding kinetics were determined using stopped-flow techniques. Dissociation rates were measured using dilution methods. GS-9256 and vedroprevir had measured Ki values of 89 pM and 410 pM, respectively, against genotype 1b NS3 protease; Ki values were higher against genotype 2a (2.8 nM and 39 nM) and genotype 3 proteases (104 nM and 319 nM) for GS-9256 and vedroprevir, respectively. Selectivity of GS-9256 and vedroprevir was >10,000-fold against all tested off-target proteases. Association rate constants of 4×10(5)M(-1)s(-1) and 1×10(6)M(-1)s(-1), respectively, were measured, and dissociation rate constants of 4.8×10(-5)s(-1) and 2.6×10(-4)s(-1) were determined. GS-9256 and vedroprevir are potent inhibitors of NS3 protease with high selectivity against off-target proteases. They have rapid association kinetics and slow dissociation kinetics. The NS3 protease is a key drug target for the treatment of hepatitis C. The potency, selectivity, and binding kinetics of GS-9256 and vedroprevir constitute a biochemical profile that supports the evaluation of these compounds in combination with other direct-acting antivirals in clinical trials for hepatitis C.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Urea, European Pharmacopoeia (EP) Reference Standard
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Supelco
Urea, analytical standard
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)