Merck
GSK-3β-regulated N-acetyltransferase 10 is involved in colorectal cancer invasion.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-07-02)
Hong Zhang, Wei Hou, Hua-Li Wang, Hai-Jing Liu, Xin-Ying Jia, Xing-Zheng Zheng, Yong-Xin Zou, Xin Li, Lin Hou, Michael A McNutt, Bo Zhang
ABSTRAKT
NAT10 (N-acetyltransferase 10) is a nucleolar protein, but may show subcellular redistribution in colorectal carcinoma. In this study, we evaluated membranous staining of NAT10 in colorectal carcinoma and its clinical implications, and explored the mechanism of regulation of NAT10 redistribution. The expression and subcellular redistribution of NAT10, β-catenin, E-cadherin, and GSK-3β were evaluated by immunohistochemistry in 222 cases of colorectal carcinoma. Regulation of NAT10 and its influence on cell motility were analyzed with inhibitors of GSK-3β, transfection of wild-type or kinase-inactivated GSK-3β, or expression of various domains of NAT10, and evaluated with immunofluorescence, Western blotting, and Transwell assays. NAT10 localized mainly in the nucleoli of normal tissues, and was redistributed to the membrane in cancer cells, particularly at the invasive "leading edge" of the tumor. This correlated well with nuclear accumulation of β-catenin (P<0.001; χ2=68.213). In addition, NAT10 membrane staining reflected the depth of invasion and tendency to metastasize (all P values<0.001), and was associated with a poorer prognosis (P=0.023; χ2=5.161). Evaluation of the mechanism involved demonstrated that subcellular redistribution of NAT10 may result from its increased stability and nuclear export, which is brought about by inhibition of GSK-3β. Moreover, redistribution of NAT10 induces alteration of cytoskeletal dynamics and increases cancer cell motility. The subcellular redistribution of NAT10 can be induced by decreases in GSK-3β activity. This redistribution increases cancer cell motility, and is, thus, correlated with invasive potential and poorer clinical outcome. This finding suggests that NAT10 may be a useful prognostic marker and potential therapeutic target in colorectal carcinoma.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Lithium chloride, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Lithium chloride, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Supelco
Lithium chloride solution, 1 M in ethanol
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
SAFC
Sodium chloride solution, 5 M