Merck
2-bromo-4-methylphenol, a compound responsible for iodine off-flavor in wines.
Journal of agricultural and food chemistry (2014-11-21)
Jean-Christophe Barbe, Sophie Tempere, Laurent Riquier, Georgia Lytra, Stéphanie Marchand, Gilles de Revel
ABSTRAKT
This study focuses on an off-flavor in wines treated by an acidification technique involving ion-exchange resins. Applying reversed-phase HPLC on a C18 column to a contaminated wine extract resulted in 25 fractions in dilute alcohol medium, and only one, fraction 19, presented this off-flavor. Its composition was analyzed using gas chromatography coupled to olfactometry (GC-O) and mass spectrometry (GC-MS), leading to the identification of 2-bromo-4-methylphenol for the first time as a compound associated with an "iodine" aroma in white, rosé, and red table wines. Contaminated wines presented a wide range of levels, with maximum concentrations of 28900, 7556, and 48000 ng/L and minimum values of 260, 35, and 63 ng/L in white, rosé, and red wines, respectively. Sensory studies investigating 2-bromo-4-methylphenol gave very interesting results. Individual thresholds ranged from 0.15 to >77.4 ng/L in reverse osmosis water, from 0.4 to >193.5 ng/L in white wine, from 0.5 to >774 ng/L in rosé wine, and from 0.5 to >774 ng/L in red wine. The very wide distribution of individual thresholds suggested the presence of two populations, sensitive and nonsensitive, with a bimodal frequency distribution. The odor descriptions quoted by the more sensitive group characterized 2-bromo-4-methylphenol using terms with "iodine" connotations in both the reverse osmosis water and red wine matrices. The descriptors for commercial wines naturally affected by this defect were seafood, iodine, mud, and fish.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Divinylbenzene, technical grade, 80%
Sigma-Aldrich
Divinylbenzene, technical grade, 55%
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material