Merck
  • A laboratory test based on determination of cytokine profiles: a promising assay to identify exposition to contact allergens and predict the clinical outcome in occupational allergic contact dermatitis.
A laboratory test based on determination of cytokine profiles: a promising assay to identify exposition to contact allergens and predict the clinical outcome in occupational allergic contact dermatitis.
BMC immunology (2015-02-06)
Valentina Bordignon, Francesca Palamara, Giorgia Altomonte, Isabella Sperduti, Mario Pietravalle, Claudia Cavallotti, Paola Cordiali-Fei, Maria Pia Fuggetta, Antonio Cristaudo, Fabrizio Ensoli
ABSTRAKT
Para-phenylenediamine (PPD) is the main allergen causing adverse reactions to hair dyes and a frequent cause of occupational-related skin sensitization among hairdressers and beauticians. The immunologic mechanism of the disease relies on the production of inflammatory cytokines by allergen-specific T cells, while regulatory T cells are thought to down-modulate the allergic response. This study was aimed at investigating the expression of effector or regulatory cytokines in exposed subjects in order to verify whether different cytokine profiles might predict distinct clinical outcomes. Peripheral blood mononuclear cells (PBMC) from 21 subjects occupationally exposed or not (10) to PPD were kept in short term cultures in the presence of optimized concentrations of NiSO4 × 6H2O or PPD. The production of IFN-γ and IL-10 elicited by antigens were analyzed by the ELISpot assay. The presence of IFN-γ responses toward PPD was significantly correlated with a positive patch test (P = 0.002) and allergic symptoms, while IL10 responses were invariably found in PPD-exposed but clinically asymptomatic subjects with negative patch testing. We found concordance between the different cytokine profiles and patch test results. No false-positive results were found for the different cytokine profiles induced by PPD, resulting in 100% specificity. The sensitivity of the test was 87.5% (95% CI 65.9-100.0) with an overall test accuracy of 93.3%. Although larger prospective-retrospective studies are necessary to validate the predictive potential of the test, the negative and positive predicted values for PPD in this study were NPV = 87.5% and PPV = 100%, respectively. These data indicate that distinct cytokine profiles are associated with different clinical manifestations. The test, which is based on a simple and rapid profiling of cytokine responses by T lymphocytes against allergens, has proven to be a promising laboratory tool, useful for both the identification of previous contact with allergens and the etiologic diagnosis of contact allergies as well as capable of predicting the clinical outcome (development of an allergic or tolerant response).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Nickel(II) sulfate hexahydrate, ReagentPlus®, powder or crystals
Sigma-Aldrich
Nickel(II) sulfate hexahydrate, ACS reagent, ≥98%
Supelco
Nickel(II) sulfate hexahydrate, ≥99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
SAFC
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%
Sigma-Aldrich
L-Glutamine
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%