Merck
Chloroquine inhibits MGC803 gastric cancer cell migration via the Toll-like receptor 9/nuclear factor kappa B signaling pathway.
Molecular medicine reports (2014-11-06)
Yanli Zhang, Yunhong Li, Yalan Li, Rui Li, Yajun Ma, Hao Wang, Yin Wang
ABSTRAKT
Stimulation of Toll‑like receptor 9 (TLR9) has been associated with invasion in various types of cancer cell in vitro. The present study aimed to evaluate the expression of TLR9 in MGC803 gastric cancer cells and investigate the effect of a non‑specific TLR9 inhibitor, chloroquine (CQ), on MGC803 cell migration via the TLR9/nuclear factor kappa B (NFκB) signaling pathway. The expression of TLR9 was investigated using reverse transcription polymerase chain reaction (RT‑PCR), flow cytometry and western blot analysis. The effects of CQ on MGC803 cell proliferation were measured by MTT colorimetric assay. The mRNA expression levels of cyclooxygenase‑2 (COX‑2), matrix metalloproteinase (MMP)‑2, MMP‑7 and NFκB p65 were evaluated by RT‑PCR in MGC803 cells stimulated by various concentrations of CQ. The migration of gastric cancer cells treated with CQ at 12, 24 and 36 h was measured by wound healing assay. The results indicated that MGC803 cells expressed TLR9 and that CQ had anti‑proliferative effects on MGC803 cells and inhibited mRNA expression of COX‑2, MMP‑2, MMP‑7 and NFκB p65 (P<0.05). Furthermore, CQ inhibited the bioactivity of NFκB p65 and prevented the migration of MGC803 cells in a dose‑dependent manner (P<0.05). In conclusion, the results indicated that the TLR9/NFκB signaling pathway was involved in gastric cancer cell migration and that CQ had anti‑tumor activity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
SAFC
BIS-TRIS
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)