Merck
R-spondin1 regulates cell proliferation of corneal endothelial cells via the Wnt3a/β-catenin pathway.
Investigative ophthalmology & visual science (2014-10-04)
Naoki Okumura, Takahiro Nakamura, EunDuck P Kay, Makiko Nakahara, Shigeru Kinoshita, Noriko Koizumi
ABSTRAKT
To evaluate the effect of Roof plate-specific spondin 1 (R-spondin1) on the proliferation of corneal endothelial cells (CECs) and to determine whether the Wnt/β-catenin pathway is involved in the activities of R-spondin1. The proliferation of rabbit CECs (RCECs) and human CECs (HCECs) was measured by 5-bromo-2'-deoxyuridine (BrdU) incorporation into DNA. The effect of R-spondin1 on CEC density was evaluated in ex vivo organ-cultured rabbit and human corneal tissues. The cell density of HCECs cultured with R-spondin1 was also evaluated in vitro. The subcellular localization of function-associated markers of CECs (zona occludens 1 [ZO-1] and Na+/K+-ATPase) was determined by immunohistocytochemistry. The expression of cell cycle proteins and localization of β-catenin were determined by immunoblotting. The in vitro proliferation of RCECs and HCECs increased by 1.2- to 1.3-fold in response to R-spondin1. The CEC densities of rabbit and human corneal tissues were increased significantly by R-spondin1 treatment. Na+/K+-ATPase and ZO-1 were well preserved on the plasma membranes. When HCECs were maintained in the presence of R-spondin1 for up to 90 days, the maximum cell density was observed at approximately 50 days, and the cell density was maintained for up to 90 days. R-spondin1 facilitated the nuclear import of β-catenin in RCECs within 30 minutes, which subsequently upregulated cyclin D and downregulated p27, leading to G1/S progression by hyperphosphorylation of the retinoblastoma protein. The unique effects of R-spondin1 on the proliferation of CECs, regardless of species, indicate that R-spondin1 may play a key role in maintaining corneal endothelium homeostasis through the Wnt/β-catenin pathway.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Supelco
Electrolyte solution, nonaqueous, 2 M LiCl in ethanol
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Supelco
Roztwór elektrolitu, nonaqueous, LiCl in ethanol (saturated)
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Kwas askorbinowy, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
SAFC
L-Threonine
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Supelco
Lithium chloride solution, 1 M in ethanol