Merck
  • Development of disulfiram-loaded poly(lactic-co-glycolic acid) wafers for the localised treatment of glioblastoma multiforme: a comparison of manufacturing techniques.
Development of disulfiram-loaded poly(lactic-co-glycolic acid) wafers for the localised treatment of glioblastoma multiforme: a comparison of manufacturing techniques.
Journal of pharmaceutical sciences (2014-12-30)
Iwona Zembko, Iram Ahmed, Aneesa Farooq, Jagdeep Dail, Patrica Tawari, Weiguang Wang, Christopher Mcconville
ABSTRAKT
Glioblastoma multiforme (GBM) is the most common primary malignant brain tumour in adults with a very poor prognosis. This paper describes the development of disulfiram (DSF)-loaded biodegradable wafers manufactured using three standard techniques: compression, solvent casting and heat compression moulding. The paper demonstrates that neither technique has an adverse effect on the stability of the DSF within the wafers. However, the solvent casting technique results in an interaction between the poly(lactic-co-glycolic acid) (PLGA) and the DSF. The physical state of the DSF within the wafers was dependent on the manufacturing technique, with the DSF in the wafers manufactured by compression or solvent casting retaining between 40% and 98% crystallinity, whereas the DSF in the wafers manufactured using heat compression moulding was completely amorphous. Release of DSF from the wafers is dependent on the degradation of the PLGA, the manufacturing technique used, and the DSF loading. DSF in the compressed and heat compression moulded wafers had a similar cytotoxicity against a GBM cell line compared with the unprocessed DSF control. However, the cytotoxicity of the DSF in the solvent-casted wafers was significantly lower than the unprocessed DSF.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
cis-11-Methyl-2-dodecenoic acid, ≥90.0% (HPLC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycolide, ≥99%
Sigma-Aldrich
3,6-Dimethyl-1,4-dioxane-2,5-dione, 99%