Merck
  • A Humanized Monoclonal Antibody against Heat Shock Protein 60 Suppresses Murine Arthritis and Colitis and Skews the Cytokine Balance toward an Anti-Inflammatory Response.
A Humanized Monoclonal Antibody against Heat Shock Protein 60 Suppresses Murine Arthritis and Colitis and Skews the Cytokine Balance toward an Anti-Inflammatory Response.
Journal of immunology (Baltimore, Md. : 1950) (2015-04-24)
Rina Ulmansky, Dorit Landstein, Eli Moallem, Virginie Loeb, Avi Levin, Ronit Meyuhas, Galia Katzavian, Shira Yair, Yaakov Naparstek
ABSTRAKT
We have previously shown that naturally occurring as well as acquired Abs against the Mycobacterium tuberculosis heat shock protein (HSP)65 protect against the induction of murine autoimmune inflammatory arthritis. In the present work, we have studied the anti-inflammatory effect of prozumab, a humanized anti-HSP mAb in murine inflammatory arthritis and colitis, and its effects on cytokine secretion. Prozumab was shown to bind to HSP60, the highly conserved mammalian homolog of the bacterial protein, and it was found to be effective in protecting and suppressing autoimmune arthritis in the models of adjuvant arthritis and collagen-induced arthritis in rats and mice, respectively, as well as in acute hapten-mediated colitis and chronic, spontaneous colitis models. Mechanistically, prozumab induces IL-10 secretion from naive human PBMCs and suppresses the secretion of IFN-γ and IL-6 from anti-CD3-activated human PBMCs. These findings make prozumab a promising potential drug for treating human rheumatoid arthritis and inflammatory bowel disease, as well as a wide range of autoimmune inflammatory diseases.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
SAFC
L-Glutamine
Sigma-Aldrich
Picrylsulfonic acid solution, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source