Merck
  • Ovarian cancer immunotherapy using PD-L1 siRNA targeted delivery from folic acid-functionalized polyethylenimine: strategies to enhance T cell killing.
Ovarian cancer immunotherapy using PD-L1 siRNA targeted delivery from folic acid-functionalized polyethylenimine: strategies to enhance T cell killing.
Advanced healthcare materials (2015-04-14)
Pei Yun Teo, Chuan Yang, Lynsey M Whilding, Ana C Parente-Pereira, John Maher, Andrew J T George, James L Hedrick, Yi Yan Yang, Sadaf Ghaem-Maghami
ABSTRAKT
Adoptive T cell immunotherapy is a promising treatment strategy for epithelial ovarian cancer (EOC). However, programmed death ligand-1 (PD-L1), highly expressed on EOC cells, interacts with programmed death-1 (PD-1), expressed on T cells, causing immunosuppression. This study aims to block PD-1/PD-L1 interactions by delivering PD-L1 siRNA, using various folic acid (FA)-functionalized polyethylenimine (PEI) polymers, to SKOV-3-Luc EOC cells, and investigate the sensitization of the EOC cells to T cell killing. To enhance siRNA uptake into EOC cells, which over express folate receptors, PEI is modified with FA or PEG-FA so that siRNA is complexed into nanoparticles with folate molecules on the surface. PEI modification with a single functional group lowers the polymer cytotoxicity compared to unmodified PEI. FA-conjugated polymers increase siRNA uptake into SKOV-3-luc cells and decrease unspecific uptake into monocytes. All polymers result in 40% to 50% PD-L1 protein knockdown. Importantly, SKOV-3-Luc cells treated with either PEI-FA or PEI- polyethylene glycol (PEG)-FA/PD-L1 siRNA complexes are up to twofold more sensitive to T cell killing compared to scrambled siRNA treated controls. These findings are the first to demonstrate that PD-L1 knockdown in EOC cells, via siRNA/FA-targeted delivery, are able to sensitize cancer cells to T cell killing.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets