Merck
Reactivity of rat bone marrow-derived macrophages to neurotransmitter stimulation in the context of collagen II-induced arthritis.
Arthritis research & therapy (2015-06-25)
Dominique Muschter, Claudia Göttl, Mandy Vogel, Joachim Grifka, Rainer H Straub, Susanne Grässel
ABSTRAKT
Numerous observations indicate that rheumatoid arthritis (RA) has a bone marrow component. In parallel, local synovial changes depend on neuronal components of the peripheral sympathetic nervous system. Here, we wanted to analyze whether collagen II-induced arthritis (CIA) has an impact on number, adhesion, apoptosis, and proliferation of the macrophage subset of bone marrow cells and how alterations in neurotransmitter microenvironment affect these properties. Bone marrow-derived macrophages (BMMs) were isolated from Dark Agouti rats at different stages of CIA, and number, adhesion, caspase 3/7 activity, and proliferation were analyzed in the presence of acetylcholine (ACh), noradrenaline (NA), and vasoactive intestinal peptide (VIP). Opposed to enhanced CD11b(+) (cluster of differentiation 11b-positive) and EMR1(+) (epidermal growth factor-like module-containing mucin-like hormone receptor-like 1-positive) cells, characterizing the macrophage subset, in native bone marrow of rats with acute inflammatory arthritis, we found decreased numbers of CIA macrophages after enrichment and culture in comparison with healthy (control) animals. Adhesion studies revealed significantly reduced attachment to plastic in acute arthritis and collagen type I and fibronectin in chronic arthritis. Additionally, we found a strong reduction in proliferation of BMMs at CIA onset and in the chronic phase of CIA. Apoptosis remained unaffected. Neurotransmitter stimulation profoundly affected proliferation, adhesion, and apoptosis of BMMs from CIA and control rats, depending on disease time point. Cultured BMMs from CIA and control animals expressed neurotransmitter receptors for ACh, VIP and NA, but the expression profile seemed not to be affected by CIA. Induction of CIA distinctly inhibits proliferation of BMMs in low- and non-inflammatory phases and reduces attachment to plastic at the acute inflammatory arthritis stage and adhesion to collagen I and fibronectin at the chronic stage. Influence of neurotransmitter stimulation on adhesion, apoptosis, and proliferation is altered by CIA depending on disease stage. We suggest an altered reactivity of BMMs to neurotransmitter stimulation caused by CIA and maybe also by aging.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Vasoactive Intestinal Peptide human, porcine, rat, ≥95% (HPLC), powder
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, BioUltra, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, ≥99% (HPLC)
Sigma-Aldrich
Acetylcholine bromide, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Vasoactive Intestinal Peptide human, porcine, rat, synthetic, ≥95% (HPLC), powder
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)