Merck
Efficient endodermal induction of human adipose stem cells using various concentrations of Activin A for hepatic differentiation.
Biochemical and biophysical research communications (2015-07-26)
Sang Woo Lee, Seon Ok Min, Seon Young Bak, Ho Kyoung Hwang, Kyung Sik Kim
ABSTRAKT
Activin A, which is a signaling molecule similar to Nodal, rapidly promotes endoderm induction of both embryonic stem (ES) cells and MSCs. Protocols for hepatic induction exhibit differences in efficiency and reproducibility depending on the specific methods or sources of MSCs. We characterized the effects of Activin A concentration on induction efficiency during hepatic differentiation of MSCs. Human MSCs (hMSCs) were differentiated into a hepatic lineage via a three-step protocol. Cells were first cultured in fetal bovine serum-free MSCs conditioned medium supplemented with Activin A (20, 50, or 100 ng/mL) for 3 days followed by treatment with additional agents. RT-PCR analysis, immunocytochemistry assays, periodic acid and Schiff's solution staining, and ELISAs were performed to confirm hepatic induction of hMSCs. Expression of genes related to the primitive foregut endoderm was observed in cells treated with higher concentration of Activin A. Gene expression related to functional primitive hepatocytes was observed in the early phases of hepatic differentiation. During the early period of the differentiation protocol, greater albumin secretion was observed when cells were treated with higher concentrations of Activin A. Thus, Activin A concentration affects the rate of endoderm induction of hMSCs, and at higher concentrations in vitro.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Argon-40Ar, 99.95 atom %
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)