Merck
  • Dual κ-agonist/μ-antagonist opioid receptor modulation reduces levodopa-induced dyskinesia and corrects dysregulated striatal changes in the nonhuman primate model of Parkinson disease.
Dual κ-agonist/μ-antagonist opioid receptor modulation reduces levodopa-induced dyskinesia and corrects dysregulated striatal changes in the nonhuman primate model of Parkinson disease.
Annals of neurology (2015-03-31)
Lisa F Potts, Eun S Park, Jong-Min Woo, Bhagya L Dyavar Shetty, Arun Singh, Steven P Braithwaite, Michael Voronkov, Stella M Papa, M Maral Mouradian
ABSTRAKT
Effective medical management of levodopa-induced dyskinesia (LID) remains an unmet need for patients with Parkinson disease (PD). Changes in opioid transmission in the basal ganglia associated with LID suggest a therapeutic opportunity. Here we determined the impact of modulating both mu and kappa opioid receptor signaling using the mixed agonist/antagonist analgesic nalbuphine in reducing LID and its molecular markers in the nonhuman primate model. 1-Methyl-4-phenyl-1,2,3,6-tetrahydropyridine-treated macaques with advanced parkinsonism and reproducible LID received a range of nalbuphine doses or saline subcutaneously as: (1) monotherapy, (2) acute coadministration with levodopa, and (3) chronic coadministration for 1 month. Animals were assessed by blinded examiners for motor disability and LID severity using standardized rating scales. Plasma levodopa levels were determined with and without nalbuphine, and postmortem brain samples were subjected to Western blot analyses. Nalbuphine reduced LID in a dose-dependent manner by 48% (p < 0.001) without compromising the anti-PD effect of levodopa or changing plasma levodopa levels. There was no tolerance to the anti-LID effect of nalbuphine given chronically. Nalbuphine coadministered with levodopa was well tolerated and did not cause sedation. Nalbuphine monotherapy had no effect on motor disability. Striatal tissue analyses showed that nalbuphine cotherapy blocks several molecular correlates of LID, including overexpression of ΔFosB, prodynorphin, dynorphin A, cyclin-dependent kinase 5, and increased phosphorylation of DARPP-32 at threonine-34. Nalbuphine reverses the molecular milieu in the striatum associated with LID and is a safe and effective anti-LID agent in the primate model of PD. These findings support repurposing this analgesic for the treatment of LID.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Tetraethylammonium borohydride, technical, ≥95% (T)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)