Merck
  • Hepatic differentiation of human adipose tissue-derived mesenchymal stem cells and adverse effects of arsanilic acid and acetaminophen during in vitro hepatic developmental stage.
Hepatic differentiation of human adipose tissue-derived mesenchymal stem cells and adverse effects of arsanilic acid and acetaminophen during in vitro hepatic developmental stage.
Cell biology and toxicology (2015-04-22)
Mi-Jeong Kwon, Seok-Jin Kang, Young-Il Park, Yool-Hee Yang, Sa-Ik Bang, Yong Ho Park, ByungJae So, Myung-Haing Cho, Hwan-Goo Kang
ABSTRAKT
In the present study, we differentiated hepatocyte-like cells (HLCs) from human adipose tissue-derived mesenchymal stem cells (AT-MSCs). The hepatic differentiation was confirmed by increases in hepatic proteins or genes, the cytochrome P450 (CYP) activities, albumin secretion, and glycogen storage. To determine the developmental toxic effect of arsanilic acid (Ars) and acetaminophen (AAP) on the hepatic development, the differentiating cells were treated with the test chemicals (below IC12.5) from day 4 to day 13. The enzymatic activities of lactate dehydrogenase (LDH), alanine aminotransferase (ALT), and aspartate aminotransferase (AST) did not significantly differ in response to Ars treatment. AAP treatment increased the activities of all enzymes in a dose-dependent manner, significantly at concentrations of 2.5 and 5 mM of AAP. On the expressions of hepatic genes for Ars, the expressions were significantly inhibited by more than 0.5 mM for Albumin (ALB), but only 2.5 mM for α-feto protein (AFP). In the AAP-treated group, the expressions of ALB and AFP were significantly decreased at the concentrations exceeding 0.625 mM. The activities of CYP3A4 were not changed by both treatments. The activities of CYP1A2 were increased by AAP, whereas it was decreased by Ars treatment. In conclusion, AAP could cause serious adverse effects during the hepatic development as compared to Ars.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
SAFC
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%