Merck
Role of contraction duration in inducing fast-to-slow contractile and metabolic protein and functional changes in engineered muscle.
Journal of cellular physiology (2015-04-11)
Alastair Khodabukus, Leslie M Baehr, Sue C Bodine, Keith Baar
ABSTRAKT
The role of factors such as frequency, contraction duration and active time in the adaptation to chronic low-frequency electrical stimulation (CLFS) is widely disputed. In this study we explore the ability of contraction duration (0.6, 6, 60, and 600 sec) to induce a fast-to-slow shift in engineered muscle while using a stimulation frequency of 10 Hz and keeping active time constant at 60%. We found that all contraction durations induced similar slowing of time-to-peak tension. Despite similar increases in total myosin heavy (MHC) levels with stimulation, increasing contraction duration resulted in progressive decreases in total fast myosin. With contraction durations of 60 and 600 sec, MHC IIx levels decreased and MHC IIa levels increased. All contraction durations resulted in fast-to-slow shifts in TnT and TnC but increased both fast and slow TnI levels. Half-relaxation slowed to a greater extent with contraction durations of 60 and 600 sec despite similar changes in the calcium sequestering proteins calsequestrin and parvalbumin and the calcium uptake protein SERCA. All CLFS groups resulted in greater fatigue resistance than control. Similar increases in GLUT4, mitochondrial enzymes (SDH and ATPsynthase), the fatty acid transporter CPT-1, and the metabolic regulators PGC-1α and MEF2 were found with all contraction durations. However, the mitochondrial enzymes cytochrome C and citrate synthase were increased to greater levels with contraction durations of 60 and 600 sec. These results demonstrate that contraction duration plays a pivotal role in dictating the level of CLFS-induced contractile and metabolic adaptations in tissue-engineered skeletal muscle.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis