Merck
Impact of Roasting on Identification of Hazelnut (Corylus avellana L.) Origin: A Chemometric Approach.
Journal of agricultural and food chemistry (2015-08-01)
Monica Locatelli, Jean Daniel Coïsson, Fabiano Travaglia, Matteo Bordiga, Marco Arlorio
ABSTRAKT
Hazelnuts belonging to different cultivars or cultivated in different geographic areas can be differentiated by their chemical profile; however, the roasting process may affect the composition of raw hazelnuts, thus compromising the possibility to identify their origin in processed foods. In this work, we characterized raw and roasted hazelnuts (Tonda Gentile Trilobata, TGT, from Italy and from Chile, Tonda di Giffoni from Italy, and Tombul from Turkey), as well as hazelnuts isolated from commercial products, with the aim to discriminate their cultivar and origin. The chemometric evaluation of selected chemical parameters (proximate composition, fatty acids, total polyphenols, antioxidant activity, and protein fingerprint by SDS-PAGE) permitted us to identify hazelnuts belonging to different cultivars and, concerning TGT samples, their different geographic origin. Also commercial samples containing Piedmontese TGT hazelnuts were correctly assigned to TGT cluster. In conclusion, even if the roasting process modifies the composition of roasted hazelnuts, this preliminary model study suggests that the identification of their origin is still possible.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Myricetin, ≥96.0%, crystalline
Sigma-Aldrich
Myricetin, ≥96.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)