Merck
  • Chronic Hypoxia-Induced Autophagy Aggravates the Neuropathology of Alzheimer's Disease through AMPK-mTOR Signaling in the APPSwe/PS1dE9 Mouse Model.
Chronic Hypoxia-Induced Autophagy Aggravates the Neuropathology of Alzheimer's Disease through AMPK-mTOR Signaling in the APPSwe/PS1dE9 Mouse Model.
Journal of Alzheimer's disease : JAD (2015-10-08)
Hui Liu, Hongyan Qiu, Qian Xiao, Weidong Le
ABSTRAKT
Alzheimer's disease (AD) is the most common form of dementia with the accumulation of senile plaques and neurofibrillary tangles in the brain. Autophagy is the key machinery for mammalian cells to degrade damaged organelles and abnormal proteins. Enormous evidence suggests that the autophagy pathway is impaired in AD. Our previous study revealed that hypoxia induced autophagic activation leading to more amyloid-β production in vitro. In this study, we investigated whether autophagic dysfunction is involved in the hypoxia mediated-pathogenesis of AD. We used APPSwe/PS1dE9 transgenic (Tg) mice and wildtype (Wt) littermates. We documented that chronic hypoxia caused more and larger senile plaques in the brains of Tg mice. In addition, chronic hypoxia induced activation of autophagy in the brains of both Wt and Tg mice, and compared to the normal autophagic flux in Wt mice, the autophagic flux was impaired in the brains of H-Tg mice with a large amount of autophagic vacuole accumulation and significant high level of P62. In an in vitro study, we showed that hypoxia-induced autophagy significantly elevated the level of hAβ42. Furthermore, we found that chronic hypoxia activated AMPK and further inhibited the mTOR signaling pathway, while inhibition of AMPK attenuated autophagy induction through the enhancement of mTOR phosphorylation. In short, our study provides new insight into the mechanism underlying chronic hypoxia-mediated AD pathogenesis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
5,5-Dimethylhydantoin, 97%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)