Merck
  • Resveratrol analog piceatannol restores the palmitic acid-induced impairment of insulin signaling and production of endothelial nitric oxide via activation of anti-inflammatory and antioxidative heme oxygenase-1 in human endothelial cells.
Resveratrol analog piceatannol restores the palmitic acid-induced impairment of insulin signaling and production of endothelial nitric oxide via activation of anti-inflammatory and antioxidative heme oxygenase-1 in human endothelial cells.
Molecular medicine reports (2015-03-31)
Sun-Oh Jeong, Yong Son, Ju Hwan Lee, Yong-Kwan Cheong, Seong Hoon Park, Hun-Taeg Chung, Hyun-Ock Pae
ABSTRAKT
Growing evidence suggests that the elevation of free fatty acids, including palmitic acid (PA), are associated with inflammation and oxidative stress, which may be involved in endothelial dysfunction, characterized by the reduced bioavailability of nitric oxide (NO) synthesized from endothelial NO synthase (eNOS). Heme oxygenase-1 (HO-1) is important in the preservation of NO bioavailability. Piceatannol (Pic), with similar chemical structure to resveratrol, is suggested to possess similar protective effects as resveratrol. In the present study, human umbilical vein endothelial cells (HUVECs), stimulated with PA, were used to examine the endothelial protective effects of Pic. Pic increased the expression of HO-1 via nuclear factor erythroid-2-related factor-2 activation in the HUVECs, and decreased the PA-induced secretions of interleukin-6 and tumor necrosis factor-α, and the formation of reactive oxygen species ROS via inhibition of NF-κB activation. Notably, following inhibition of HO-1 activity by tin protoporphryin-IX, Pic did not prevent cytokine secretion, ROS formation, and NF-κB activation in the PA-stimulated HUVECs. PA attenuated insulin-mediated insulin receptor substrate-1 (IRS-1) tyrosine phosphorylation, leading to decreased glucose uptake, and phosphorylation of eNOS, leading to a reduction in the production of NO. Pic effectively mitigated the inhibitory effects of PA on the insulin-mediated phosphorylation of IRS-1 and eNOS, which was not observed following inhibition of HO‑1 activity. The results of the present study suggested that Pic may have the potential to prevent PA-induced impairment of insulin signaling and eNOS function, by inducing the expression of the anti-inflammatory and antioxidant, HO-1.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DAF-FM, ≥98% (HPLC)
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Nfe2l2
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Informacje o cenach i dostępności nie są obecnie dostępne.