Merck
  • Analysis of Glycogen Synthase Kinase Inhibitors That Regulate Cytochrome P450 Expression in Primary Human Hepatocytes by Activation of β-Catenin, Aryl Hydrocarbon Receptor and Pregnane X Receptor Signaling.
Analysis of Glycogen Synthase Kinase Inhibitors That Regulate Cytochrome P450 Expression in Primary Human Hepatocytes by Activation of β-Catenin, Aryl Hydrocarbon Receptor and Pregnane X Receptor Signaling.
Toxicological sciences : an official journal of the Society of Toxicology (2015-08-12)
Philippe Briolotti, Laurent Chaloin, Patrick Balaguer, Franck Da Silva, Veronika Tománková, Jean-Marc Pascussi, Cédric Duret, Jean-Michel Fabre, Jeanne Ramos, Sylvie Klieber, Patrick Maurel, Martine Daujat-Chavanieu, Sabine Gerbal-Chaloin
ABSTRAKT
Cytochrome P450 (CYP) expression and activity are not homogeneous in the liver lobules. Indeed, CYPs are mainly expressed and induced in centrilobular hepatocytes. The wingless-type MMTV integration site family (WNT)/β-catenin pathway was identified as a major regulator of this zonal organization. We have recently demonstrated that in primary human hepatocytes (PHHs), the expression of CYP2E1, CYP1A2, and aryl hydrocarbon receptor (AhR), but not of CYP3A4, is regulated by the WNT/β-catenin pathway in response to WNT3a, its canonical activator. Here, we investigated whether glycogen synthase kinase 3β (GSK3β) inhibitors, which mimic the action of WNT molecules, could be used in PHHs to activate the β-catenin pathway to study CYP expression. We assessed the activity of 6BIO (6-bromoindirubin-3'-oxime), CHIR99021 (6-((2-((4-(2,4-dichlorophenyl)-5-(4methyl-1H-imidazol-2-yl)pyrimidin-2-yl)amino)ethyl)amino) nicotinonitrile), and GSK3iXV (Pyridocarbazolo-cyclopentadienyl Ruthenium complex GSK3 inhibitor XV) that belong to structurally different families of GSK3β inhibitors. Using small interfering RNAs, reporter gene assays, and molecular docking predictions, we demonstrated that GSK3β inhibitors can activate the WNT/β-catenin pathway in PHHs to regulate CYP2E1 expression. We also found that 6BIO and GSK3iXV are AhR full agonists that participate, through AhR signaling, to CYP1A2 induction. Conversely, CHIR99021 is an AhR partial agonist, and a pregnane X receptor ligand and partial agonist, thus regulating CYP1A2 and CYP3A4 gene expression in a β-catenin-independent manner. In conclusion, GSK3β inhibitors can activate the WNT/β-catenin pathway in PHHs. Nevertheless, their role in CYP regulation should be analyzed with caution as these molecules can interact with xenosensors.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
BIO, ≥98% (HPLC)
Sigma-Aldrich
CH-223191
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
3-Pyridinecarbonitrile, 98%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Chlorzoxazone
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®